Merck
  • Design, synthesis and evaluation of biotin decorated inulin-based polymeric micelles as long-circulating nanocarriers for targeted drug delivery.
Design, synthesis and evaluation of biotin decorated inulin-based polymeric micelles as long-circulating nanocarriers for targeted drug delivery.
Nanomedicine : nanotechnology, biology, and medicine (2017-01-25)
Delia Mandracchia, Antonio Rosato, Adriana Trapani, Theodora Chlapanidas, Isabella Monia Montagner, Sara Perteghella, Cinzia Di Franco, Maria Luisa Torre, Giuseppe Trapani, Giuseppe Tripodo
ABSTRAKT
Here, long-circulating behaviors of Inulin-based nanomicelles are demonstrated for the first time in vivo. We show the synthesis and evaluation of biotin (BIO)-decorated polymeric INVITE micelles constituted of substances of natural origin, Inulin (INU) and Vitamin E (VITE), as long-circulating carriers for receptor-mediated targeted drug delivery. The resulting INVITE or INVITE-BIO micelles, nanometrically sized, did not reveal any cytotoxicity after 24h of incubation with Caco-2 cells. Moreover, in vitro studies on Caco-2 cells monolayers indicated that the transport of INVITE-BIO micelles was faster than surface unmodified INVITE micelles. In vivo optical imaging studies evidenced that, upon intravenous administration, INVITE-BIO micelles were quantitatively present in the body up to 48h. Instead, after oral administration, the micelles were not found in the systemic circulation but eliminated with the normal intestinal content. In conclusion, INVITE-BIO micelles may enhance drug accumulation in tumor-cells over-expressing the receptor for biotin through receptor mediated endocytosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
Sigma-Aldrich
DCC, 99%