Merck
Aerobic degradation of fenvalerate by a Gram-positive bacterium,
3 Biotech (2017-09-29)
Sikandar I Mulla, Fuad Ameen, Preeti N Tallur, Ram Naresh Bharagava, Manjunatha Bangeppagari, Syed Ali Musstjab Akber Shah Eqani, Zabin K Bagewadi, Gurumurthy D Mahadevan, Chang-Ping Yu, Harichandra Z Ninnekar
ABSTRAKT
Synthetic pyrethroid-fenvalerate-is one of the most widespread toxic pollutants and has adverse effect on living systems. However, little is known about its biotransformation mechanism in different microorganisms. To elucidate the pathway that might be involved in the catabolism of fenvalerate, we used
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Methylcatechol, 98%
Sigma-Aldrich
4-Chlorocatechol, 97%