Merck
Synthesis, characterization and biological applications of mycosynthesized silver nanoparticles.
3 Biotech (2017-09-29)
Sathiyaseelan Anbazhagan, Shajahan Azeez, Girilal Morukattu, Ramachandran Rajan, Kaviyarasan Venkatesan, Kalaichelvan Puthupalayam Thangavelu
ABSTRAKT
Silver nanoparticles (AgNPs) have been known for their inhibitory and bactericidal effects. In the present study, less toxic AgNPs using
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)