Merck
  • Galangin ameliorates cisplatin-induced nephrotoxicity by attenuating oxidative stress, inflammation and cell death in mice through inhibition of ERK and NF-kappaB signaling.
Galangin ameliorates cisplatin-induced nephrotoxicity by attenuating oxidative stress, inflammation and cell death in mice through inhibition of ERK and NF-kappaB signaling.
Toxicology and applied pharmacology (2017-06-01)
Yu-Ching Huang, Ming-Shiun Tsai, Pei-Chi Hsieh, Jheng-Hong Shih, Tsu-Shing Wang, Yi-Chun Wang, Ting-Hui Lin, Sue-Hong Wang
ABSTRAKT
Cisplatin is a chemotherapeutic agent widely used in the treatment of various cancers. However, cisplatin can induce nephrotoxicity and neurotoxicity, limiting its dosage and usage. Galangin, a natural flavonol, has been found to exhibit anti-oxidant and anti-inflammatory effects in vivo. Here, we investigated the effects of galangin on cisplatin-induced acute kidney injury (AKI) and its molecular mechanisms in mice. Galangin administration reduced the cisplatin-induced oxidative stress by decreasing renal MDA and 3-NT formations. Galangin administration also increased renal anti-oxidative enzyme activities (SOD, GPx, and CAT) and GSH levels depleted by cisplatin. Furthermore, galangin administration inactivated stress-induced Nrf2 protein and its downstream products, HO-1 and GCLC. In terms of the inflammatory response, galangin administration reduced IκBα phosphorylation, NF-κB phosphorylation and nuclear translocation, and then inhibited cisplatin-induced secretions of pro-inflammatory TNF-α, IL-1β and IL-6. In addition, cisplatin-induced ERK and p38 phosphorylations were inhibited by galangin administration. In terms of cell death, galangin administration reduced levels of p53, pro-apoptotic Bax and activated caspase-3 to inhibit the cisplatin-induced apoptosis. Galangin administration also reduced the expression levels of RIP1 and RIP3 to inhibit cisplatin-induced RIP1/RIP3-dependent necroptosis. Therefore, galangin administration significantly ameliorates cisplatin-induced nephrotoxicity by attenuating oxidative stress, inflammation, and cell death through inhibitions of ERK and NF-κB signaling pathways. Galangin might be a potential adjuvant for clinical cisplatin therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Nitro-L-tyrosine, crystalline
Sigma-Aldrich
Galangin, autophagy inducing flavonoid