Merck
Tumor antigen glycosaminoglycan modification regulates antibody-drug conjugate delivery and cytotoxicity.
Oncotarget (2017-10-06)
Helena C Christianson, Julien A Menard, Vineesh Indira Chandran, Erika Bourseau-Guilmain, Dmitry Shevela, Jon Lidfeldt, Ann-Sofie Månsson, Silvia Pastorekova, Johannes Messinger, Mattias Belting
ABSTRAKT
Aggressive cancers are characterized by hypoxia, which is a key driver of tumor development and treatment resistance. Proteins specifically expressed in the hypoxic tumor microenvironment thus represent interesting candidates for targeted drug delivery strategies. Carbonic anhydrase (CAIX) has been identified as an attractive treatment target as it is highly hypoxia specific and expressed at the cell-surface to promote cancer cell aggressiveness. Here, we find that cancer cell internalization of CAIX is negatively regulated by post-translational modification with chondroitin or heparan sulfate glycosaminoglycan chains. We show that perturbed glycosaminoglycan modification results in increased CAIX endocytosis. We hypothesized that perturbation of CAIX glycosaminoglycan conjugation may provide opportunities for enhanced drug delivery to hypoxic tumor cells. In support of this concept, pharmacological inhibition of glycosaminoglycan biosynthesis with xylosides significantly potentiated the internalization and cytotoxic activity of an antibody-drug conjugate (ADC) targeted at CAIX. Moreover, cells expressing glycosaminoglycan-deficient CAIX were significantly more sensitive to ADC treatment as compared with cells expressing wild-type CAIX. We find that inhibition of CAIX endocytosis is associated with an increased localization of glycosaminoglycan-conjugated CAIX in membrane lipid raft domains stabilized by caveolin-1 clusters. The association of CAIX with caveolin-1 was partially attenuated by acidosis,
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Heparinase III from Flavobacterium heparinum, Lyophilized powder stabilized with approx. 25% (w/w) bovine serum albumin, ≥30 units/mg protein (enzyme + BSA)
Sigma-Aldrich
MISSION® shRNA, Lentiviral Particles
Sigma-Aldrich
Heparin−Agarose, (1:1 suspension in a 20% ethanol solution)
Sigma-Aldrich
Chondroitinase ABC from Proteus vulgaris, lyophilized powder, 0.3-3 units/mg solid
Sigma-Aldrich
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline
Sigma-Aldrich
Chondroitin sulfate A sodium salt from bovine trachea, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Transduction Particles, Targets no known mammalian genes
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture