Merck
Effects of ZD0947, a novel and potent ATP-sensitive K
European journal of pharmacology (2016-10-30)
Keisuke Mori, Yoshio Yamashita, Noriyoshi Teramoto
ABSTRAKT
The effects of ZD0947, a novel ATP-sensitive K
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ZD0947, ≥98% (HPLC)