Merck
Review of titanate coupling agents and their application for dental composite fabrication.
Dental materials journal (2017-06-28)
Nidal Wanis Elshereksi, Mariyam Ghazali, Andanastuti Muchtar, Che Husna Azhari
ABSTRAKT
Silane is a dominant coupler that is widely used in dentistry to promote adhesion among the components of dental composites. Silica-based fillers can be easily silanized because of their similarly ordered structure. However, silane is hydrolytically degraded in the aqueous oral environment and inefficiently bonds to non-silica fillers. Thus, the development of hydrolytically stable dental composites is an important objective in the research on dental materials. Titanate coupling agents (TCAs) exhibit satisfactory interfacial bonding, enhanced homogeneous filler dispersion, and improved mechanical properties of the composites. Titanates also provide superior hydrolytic stability in wet environments, which should be considered in fabricating dental composites. The addition of a small amount of titanates can improve the resistance of the composites to moisture. This paper reviews the effects of the instability of silanes in moisture on the performance of dental composites and presents TCAs as alternative couplers to silanes for fabricating dental composites.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)