Merck
  • Protective effects of lipoic acid against acrylamide-induced neurotoxicity: involvement of mitochondrial energy metabolism and autophagy.
Protective effects of lipoic acid against acrylamide-induced neurotoxicity: involvement of mitochondrial energy metabolism and autophagy.
Food & function (2017-11-22)
Ge Song, Zhigang Liu, Luanfeng Wang, Renjie Shi, Chuanqi Chu, Man Xiang, Qi Tian, Xuebo Liu
ABSTRAKT
Acrylamide (ACR) is a chronic neurotoxin that is generated in high-starch foods during heat processing. Alpha-lipoic acid (LA) is an antioxidant that occurs in most plants and animals. The objective of this study was to reveal the mechanism of ACR-triggered neurotoxicity and identify the protective role of LA in SH-SY5Y cells. In this study, LA restored ACR-stimulated depletion of glutathione content and mitochondrial membrane potential, moderated the activation of inflammatory pathways, and recovered the Keap1/Nrf2 pathway. Moreover, LA upregulated the activities of oxidative phosphorylation complexes and diminished ACR-induced variation in AMPK/GSK3β, Ca
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)