Merck
  • Mesoporous silica nanoparticles (MSNs)-based organic/inorganic hybrid nanocarriers loading 5-Fluorouracil for the treatment of colon cancer with improved anticancer efficacy.
Mesoporous silica nanoparticles (MSNs)-based organic/inorganic hybrid nanocarriers loading 5-Fluorouracil for the treatment of colon cancer with improved anticancer efficacy.
Colloids and surfaces. B, Biointerfaces (2017-08-19)
Gang Pan, Ting-Ting Jia, Qian-Xia Huang, Yan-Yan Qiu, Jie Xu, Pei-Hao Yin, Tao Liu
ABSTRAKT
Novel methods to improve the anticancer performance of 5-fluorouracil (5-FU) is quite necessary for clinical medicines. In the present work, we fabricated a novel type of mesoporous silica nanoparticles (MSNs)-based inorganic/organic hybrid nanoparticles covalently attached with poly(oligo(ethylene glycol) monomethyl ether methacrylate) (POEGMA) for improved stabilization and targeting peptide (RGD) for targeted delivery with the aim of improving the anticancer performance of 5-FU. Atom transfer radical polymerization (ATRP) initiator functionalized MSN (MSN-Br) was synthesized at first, which was followed by surface-initiated ATRP of water soluble OEGMA and carboxyl-containing monomer (2-succinyloxyethyl methacrylate, SEMA). Functionalization of RGD onto the hydrophilic P(OEGMA-co-SEMA) chains afforded the final hybrid nanoparticle, MSN-P(OEGMA-co-RGD). 5-FU can be effectively loaded into the meso-pores of MSN-P(OEGMA-co-RGD) (5-FU@MSN-RGD) with drug content ∼7.5wt%. And the dynamic diameter (D
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%