- One-pot synthesis of magnetic zeolitic imidazolate framework/grapheme oxide composites for the extraction of neonicotinoid insecticides from environmental water samples.
One-pot synthesis of magnetic zeolitic imidazolate framework/grapheme oxide composites for the extraction of neonicotinoid insecticides from environmental water samples.
Journal of separation science (2017-10-11)
Xiaolin Cao, Zejun Jiang, Shanshan Wang, Sihui Hong, Hui Li, Yong Shao, Yongxin She, Jing Wang, Fen Jin, Maojun Jin
PMID28988436
ABSTRAKT
Magnetic zeolitic imidazolate framework 67/graphene oxide composites were synthesized by one-pot method at room temperature for the first time. Electrostatic interactions between positively charged metal ions and both negatively charged graphene oxide and Fe
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine, PESTANAL®, analytical standard