Merck
Continuous treatment with FTS confers resistance to apoptosis and affects autophagy.
PloS one (2017-02-06)
Eran Schmukler, Eya Wolfson, Zvulun Elazar, Yoel Kloog, Ronit Pinkas-Kramarski
ABSTRAKT
High percentage of human cancers involves alteration or mutation in Ras proteins, including the most aggressive malignancies, such as lung, colon and pancreatic cancers. FTS (Salirasib) is a farnesylcysteine mimetic, which acts as a functional Ras inhibitor, and was shown to exert anti-tumorigenic effects in vitro and in vivo. Previously, we have demonstrated that short-term treatment with FTS also induces protective autophagy in several cancer cell lines. Drug resistance is frequently observed in cancer cells exposed to prolonged treatment, and is considered a major cause for therapy inefficiency. Therefore, in the present study, we examined the effect of a prolonged treatment with FTS on drug resistance of HCT-116 human colon cancer cells, and the involvement of autophagy in this process. We found that cells grown in the presence of FTS for 6 months have become resistant to FTS-induced cell growth inhibition and cell death. Furthermore, we discovered that the resistant cells exhibit altered autophagy, reduced apoptosis and changes in Ras-related signaling pathways following treatment with FTS. Moreover we found that while FTS induces an apoptosis-related cleavage of p62, the FTS-resistant cells were more resistant to apoptosis and p62 cleavage.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Kalpeptyna, A cell-permeable calpain inhibitor (ID50 = 52 nM for calpain-1; ID50 = 34 nM for calpain-2; ID50 = 138 nM for papain).
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP), meets EP testing specifications
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-S6-Kinase (pThr389) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution