- Negative inotropic effects of diadenosine tetraphosphate are mediated by protein kinase C and phosphodiesterases stimulation in the rat heart.
Negative inotropic effects of diadenosine tetraphosphate are mediated by protein kinase C and phosphodiesterases stimulation in the rat heart.
European journal of pharmacology (2017-12-14)
Nikolai Pakhomov, Ksenia Pustovit, Victoria Potekhina, Tatiana Filatova, Vladislav Kuzmin, Denis Abramochkin
PMID29233660
ABSTRAKT
Extracellular diadenosine polyphosphates (Ap