Merck
Report: Palladium glutathione, N-acetylcysteine, D-penicillamine conjugation chemistry.
Pakistan journal of pharmaceutical sciences (2018-01-20)
Muhammad Mukhtiar, Syed Umer Jan, Muhammad Farid Khan, Naseem Ullah, Abid Hussain, Saifur Rehman, Muhammad Murtaza Qureshi
ABSTRAKT
The metalloelement Palladium has a number of potential Pharmaco-clinical advantages. Palladium compounds have antiviral, antibacterial, neuroprotective and antitumor properties. However studies have also indicated some mild to serious toxic effects of Palladium metalloelements. Biothiols are important antioxidants that provide protection against metals toxicity. The interaction of metalloelements with biothiols can provide valuable information about the level of toxicity of the metalloelements and about the protective role of biothiols thereof. In this piece of work the effect of salt and complexes of Palladium on the status of different thiols (GSH, NAC, and D-Pen) in aqueous medium, were examined, The thiol quantification was carried out using Elman's method through UV-visible spectrophotometry and
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
D-Penicillamine, 98-101%