manufacturer/tradename
Benchmark Scientific BR5000-STACK
compatibility
BMSB3D5000, BMSB3D5000E, BMSB3D5000EUK
General description
Optional Stacking Platform with Flat Mat workable clearance 3.5 inches and maximum clearance of 4.5 inches.
Legal Information
Everlast is a trademark of Benchmark Scientific, Inc.
1 of 1
此商品 | |||
|---|---|---|---|
| description - | description pkg of 1 ea | description pkg of 1 ea | description pkg of 1 ea |
| manufacturer/tradename Benchmark Scientific BR5000-STACK | manufacturer/tradename Benchmark Scientific BR1000-STACK | manufacturer/tradename Benchmark Scientific | manufacturer/tradename Benchmark Scientific BR1000-STACK-D |
| compatibility BMSB3D5000, BMSB3D5000E, BMSB3D5000EUK | compatibility - | compatibility - | compatibility - |