Merck
跳至
GN780263
Greiner MASTERBLOCK®储存微孔板
96 well, polypropylene, capacity 1.0 mL, round (U) bottom, red
别名:
Greiner 780263, culture dish, culture plate, greiner plate, multiwell plate
登录并加购,请在购物车里查看协议价、货期和发货地。
选择规格
变更视图
规格/货号是否有货价格
关于此项目
UNSPSC Code:
41121800
eCl@ss:
32039090
Material:
polypropylene
Size:
96 wells
价格与库存信息目前不能提供
技术服务
需要帮助?我们经验丰富的科学家团队随时乐意为您服务。
让我们为您提供帮助
material
polypropylene
packaging
pack of 50 ea
manufacturer/tradename
Greiner 780263
availability
available only in China
size
96 wells
suitability
suitable for (general storage, liquid handling; automated applications)
Legal Information
MASTERBLOCK is a registered trademark of Greiner Bio-One GmbH
Still not finding the right product?
历史批次信息供参考:
分析证书(COA)