Nucleic Acid Modifying Enzyme Selection Chart (Ultra Pure)
DNA Polymerases (ultra pure)
DNA Polymerases synthesize DNA from nucleotides. DNA polymerase is required for DNA replication, but also essential for many other activities in the cell, including genetic recombination, DNA repair, and reverse transcription. In molecular biology labs, DNA polymerases are used for applications such as cloning, PCR, and DNA sequencing.
Each DNA polymerase has special characteristics, making it useful for specific applications. Knowing these characteristics enables you to pick the right ultra pure DNA Polymerase for your application.
DNA Polymerase Selection Chart | |||
|---|---|---|---|
| Product Number | Product Description | Notable Characteristics | Application Suitability |
| KEM0026 | Klenow (3’ → 5’ exo) |
|
|
| KEM0027 | Klenow (3’ → 5’ exo) | ||
| KEM0028 | phi29 DNA Polymerase (High concentration) |
|
|
| KEM0029 | phi29 DNA Polymerase (Low concentration) | ||
| KEM0030 | DNA Polymerase I |
|
|
| KEM0031 | Klenow Fragment |
|
|
| KEM0032 | Terminal deoxynucleotidyl Transferase (TdT) |
|
|
| KEM0033 | T4 DNA Polymerase |
|
|
| KEM0034 | Mako DNA Polymerase (3’ → 5’ exo) |
|
|
| KEM0035 | Manta 1.0 DNA Polymerase (High concentration) |
|
|
| KEM0036 | Manta 1.0 DNA Polymerase (Low concentration) | ||
DNA Ligases (ultra pure)
DNA Ligases form phosphodiester bonds between DNA strands, joining them together. DNA Ligase is responsible for repairing breaks in single and double stranded DNA. In molecular biology labs, DNA Ligase is commonly used for molecular cloning applications.
Each DNA Ligase has special characteristics, making it useful for specific applications. Knowing these characteristics enables you to pick the right ultra pure DNA Ligase for your application.
DNA Ligase Selection Chart | |||||
|---|---|---|---|---|---|
| Product Description | T4 DNA Ligase | T4 DNA Ligase (Rapid) | T3 DNA Ligase | T7 DNA Ligase | E. coli DNA Ligase |
| Products No. | KEM0020 | KEM0019 | KEM0017 | KEM0018 | KEM0025 |
| Units | 150,000 | 240,000 | 900,000 | 900,000 | 2,500 |
| Concentration | 120,000 U/mL | 600,000 U/mL | 3,000,000 U/mL | 3,000,000 U/mL | 40,000 U/mL |
| Cofactor Required | ATP | ATP | ATP | ATP | NAD |
| Recommended for Cloning? | ✓ | ✓ | |||
| Recommended for cohesive ends | ✓ | ✓ | ✓ | ✓ | ✓ |
| Recommended for blunt ends | ✓ | ✓ | ✓ | ✓ | ✓ |
| Recommended for nicks in dsDNA | ✓ | ✓ | ✓ | ✓ | ✓ |
| Recommended for joining of RNA & DNA hybrids | ✓ | ✓ | |||
| Application Suitability |
|
|
|
|
|
Nucleic Acid Modifying Enzymes (ultra pure)
Many different enzymes exist that are able to modify DNA, RNA, and RNA:DNA hybrids. In molecular biology labs, these modifying enzymes are commonly used for molecular cloning and sequencing applications.
Each enzyme has special characteristics, making it useful for specific applications. Knowing these characteristics enables you to pick the right ultra pure Nucleic Acid Modifying Enzyme for your application.
Nucleic Acid Modifying Enzyme Selection Chart | |||
|---|---|---|---|
| Product No. | Product Description | Notable Characteristics | Application Suitability |
| KEM0006 | T4 Polynucleotide Kinase |
|
|
| KEM0001 | Uracil DNA Glycosylase (UDG) |
|
|
| KEM0002 | Thermolabile Uracil DNA Glycosylase (UDG) |
| |
| KEM0004 | Exonuclease III |
|
|
| KEM0005 | Lambda Exonuclease |
|
|
| KEM0009 | End-Repair Mix (High Concentration) |
|
|
| KEM0010 | End-Repair Mix (Low Concentration) | ||
| KEM0011 | 10x Uracil Cleavage System |
|
|
| KEM0014 | Poly(A) Polymerase |
|
|
| KEM0015 | RNase H |
|
|
RNA Ligases (ultra pure)
RNA Ligases form phosphodiester bonds between RNA strands, joining them together. In molecular biology labs, RNA Ligase is commonly used for mutagenesis of RNA, and end-labeling applications.
Each RNA Ligase has special characteristics, making it useful for specific applications. Knowing these characteristics enables you to pick the right ultra pure RNA Ligase for your application.
| Product Description | T4 RNA Ligase I | T4 RNA Ligase 2 | T4 RNA Ligase 2 (truncated) |
| Product No. | KEM0021 | KEM0024 | KEM0023 |
| Units | 10,000 | 4,500 | 500 |
| Concentration | 20,000 U/mL | 30,000 U/mL | 5,000 U/mL |
| Cofactor Required | ATP | ATP | None |
| Recommended for nicks in dsDNA? | ✓ | ||
| Recommended for joining of RNA:DNA hybrids? | ✓ | ||
| Recommended for labeling of RNA 3’ termini? | ✓ | ✓ | |
| Recommended for joining ssDNA? | ✓ | ||
| Recommended for joining ssRNA? | ✓ |