Skip to Content
Merck

GF67339963

Palladium

foil, 8mm disks, thickness 0.03mm, as rolled, 99.95%

Synonym(s):

Palladium, PD000261

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
MDL number:
UNSPSC Code:
12141733
PubChem Substance ID:
NACRES:
NA.23

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 673-399-63

resistivity

9.96 μΩ-cm, 20°C

diam. × thickness

8 mm × 0.03 mm

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF44617619GF74894136GF07375170
form

foil

form

foil

form

foil

form

foil

assay

99.95%

assay

99.95%

assay

99.95%

assay

99.95%

manufacturer/tradename

Goodfellow 673-399-63

manufacturer/tradename

Goodfellow 446-176-19

manufacturer/tradename

Goodfellow 748-941-36

manufacturer/tradename

Goodfellow 073-751-70

mp

1554 °C (lit.)

mp

1554 °C (lit.)

mp

1554 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masahiro Toyota
Natural product communications, 8(7), 999-1004 (2013-08-29)
A novel palladium-catalyzed intramolecular oxidative alkylation of unactivated olefins is described. This protocol was devised to solve one of the drawbacks of the original palladium-catalyzed cycloalkenylation that we developed. We call this new procedure the 'second generation palladium-catalyzed cycloalkenylation'. This
Synthesis of heterocycles via palladium-catalyzed carbonylations.
Xiao-Feng Wu et al.
Chemical reviews, 113(1), 1-35 (2012-10-09)
Update 1 of: Synthesis and functionalization of indoles through palladium-catalyzed reactions.
Sandro Cacchi et al.
Chemical reviews, 111(5), PR215-PR283 (2011-05-12)
Amanda J Hickman et al.
Nature, 484(7393), 177-185 (2012-04-14)
Copper and palladium catalysts are critically important in numerous commercial chemical processes. Improvements in the activity, selectivity and scope of these catalysts could drastically reduce the environmental impact, and increase the sustainability, of chemical reactions. One rapidly developing strategy for
Anton V Dubrovskiy et al.
Combinatorial chemistry & high throughput screening, 15(6), 451-472 (2012-01-26)
The iodocyclization of functionally-substituted alkynes provides an excellent way to prepare a wide range of iodoheterocycles, which can then be readily elaborated through palladium-catalyzed Suzuki-Miyaura, Sonogashira, Heck, Hartwig-Buchwald, and carbonylation processes into libraries of medicinally relevant heterocycles. The synthesis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service