跳轉至內容
Merck
Strychnine poisoning: gone but not forgotten.
Emergency medicine journal : EMJ (2010-09-03)
A J Parker, J B Lee, J Redman, L Jolliffe
摘要
Strychnine was used as a pesticide until 1968 and a rodenticide until 2006 when its sale was banned throughout the EU and all supplies recalled. A case of strychnine poisoning seen in a UK emergency department in 2009 is reported to remind clinicians of the features and management of this increasingly rare presentation. Prompt recognition and early intensive supportive therapy can result in a favourable outcome.