跳轉至內容
Merck
  • Apigenin-induced apoptosis of leukemia cells is mediated by a bimodal and differentially regulated residue-specific phosphorylation of heat-shock protein-27.
Apigenin-induced apoptosis of leukemia cells is mediated by a bimodal and differentially regulated residue-specific phosphorylation of heat-shock protein-27.
Cell death & disease (2011-03-03)
M E Gonzalez-Mejia, O H Voss, E J Murnan, A I Doseff
摘要
Apigenin, a natural plant flavonoid with antiproliferative activity, is emerging as a promising compound for cancer prevention and therapy, but its mechanism of action remains unclear. High expression of the small heat-shock protein-27 (Hsp27) in leukemia contributes to the resistance of these cells to cancer treatments. Changes in Hsp27 phosphorylation have been associated with heat and metabolic stress, but its role in flavonoid anticancer activity has not been investigated. In this study, we examined the effect of apigenin in the regulation of Hsp27 on leukemia. We showed that apigenin does not affect Hsp27 expression but induces a bimodal phosphorylation on Ser78 and Ser82. The phosphorylation at early times was regulated by p38. At later times, Hsp27 phosphorylation was dependent on p38 activity and for some residues on PKCδ. Silencing of p38 expression reduced apigenin-induced phosphorylation on Ser15, Ser78, and Ser82, whereas silencing of PKCδ expression reduced the phosphorylation on Ser15 and Ser82 without affecting Ser78. In addition, we found that apigenin-induced PKCδ activity is mediated by p38. We also showed that the phosphorylation of Hsp27 significantly increased the susceptibility of leukemia cells to apigenin-induced apoptosis. Together, these results identify a complex signaling network regulating the cytotoxic effect of apigenin through Hsp27 phosphorylation.