MilliporeSigma
Skip To
WPI-FLOX-PATCH
WPI
WPI Oxygen Sensor
Non-Invasive Oxygen Monitoring Kit, including phase measurement system, temperature probe
A Trusted Marketplace Seller
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 EA
Available to ship onJuly 10, 2026
$4,741.38
$4,741.38
Available to ship onJuly 10, 2026Details
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
WPI
General description
FLOX is a device for measuring fluorescence lifetime, phase and intensity. It uses LED excitation and photodiode detection with filter-based wavelength selection for easy experimental set-up and control.
FLOX is especially useful for oxygen sensing applications where stability and sensitivity to drift is important and where sample set-ups must be left undisturbed for long periods of time.
Choose from one of the most recent versions:
Certificates of Analysis (COA)