Saltar al contenido
MilliporeSigma

Saltar a

1A08060

USP

Clomiphene Benzophenone Analog

Pharmaceutical Analytical Impurity (PAI)

Iniciar sesión para ver los precios por organización y contrato.

Seleccione un Tamaño

Cambiar Vistas
Talla/SKUDisponibilidadPrecio

Acerca de este artículo

Fórmula empírica (notación de Hill):
C19H23NO2
Número CAS:
Peso molecular:
297.39
UNSPSC Code:
41116107
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarle


grade

pharmaceutical analytical impurity (PAI)

agency

USP

form

liquid

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

O=C(C=1C=CC=CC1)C2=CC=C(OCCN(CC)CC)C=C2

InChI

InChI=1S/C19H23NO2/c1-3-20(4-2)14-15-22-18-12-10-17(11-13-18)19(21)16-8-6-5-7-9-16/h5-13H,3-4,14-15H2,1-2H3

InChI key

GSTOOYJSPRJGDO-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Find answers to frequently asked questions on USP products in USP FAQs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here.

Comparar elementos similares

Ver comparación completa

Mostrar Diferencias

1 of 1

Este artículo
1A0742011400001140101
description

Pharmaceutical Analytical Impurity (PAI)

description

Pharmaceutical Analytical Impurity (PAI)

description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

grade

pharmaceutical analytical impurity (PAI)

grade

pharmaceutical analytical impurity (PAI)

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

format

neat

format

neat

format

neat

format

neat

form

liquid

form

powder

form

-

form

-

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

agency

USP

agency

USP

agency

USP

agency

USP


Clase de almacenamiento

10 - Combustible liquids



Elija entre una de las versiones más recientes:

Certificados de análisis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documentos section.

Si necesita más asistencia, póngase en contacto con Atención al cliente

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos