MilliporeSigma
  • Changes in Intestinal Microbiota and Predicted Metabolic Pathways During Colonic Fermentation of Mango (Mangifera indica L.)-Based Bar Indigestible Fraction.
Changes in Intestinal Microbiota and Predicted Metabolic Pathways During Colonic Fermentation of Mango (Mangifera indica L.)-Based Bar Indigestible Fraction.
Nutrients (2020-03-07)
Wilbert Gutiérrez-Sarmiento, Sonia Guadalupe Sáyago-Ayerdi, Isabel Goñi, Federico Antonio Gutiérrez-Miceli, Miguel Abud-Archila, José Del Carmen Rejón-Orantes, Reiner Rincón-Rosales, Betsy Anaid Peña-Ocaña, Víctor Manuel Ruíz-Valdiviezo
RESUMEN
Mango (Mangifera indica L.) peel and pulp are a source of dietary fiber (DF) and phenolic compounds (PCs) that constituent part of the indigestible fraction (IF). This fraction reaches the colon and acts as a carbon and energy source for intestinal microbiota. The effect of mango IF on intestinal microbiota during colonic fermentation is unknown. In this study, the isolated IF of a novel 'Ataulfo' mango-based bar (snack) UV-C irradiated and non-irradiated (UVMangoB and MangoB) were fermented. Colonic fermentation occurred in vitro under chemical-enzymatic, semi-anaerobic, batch culture and controlled pH colonic conditions. Changes in the structure of fecal microbiota were analyzed by 16s rRNA gene Illumina MiSeq sequencing. The community´s functional capabilities were determined in silico. The MangoB and UVMangoB increased the presence of Faecalibacterium, Roseburia, Eubacterium, Fusicatenibacter, Holdemanella, Catenibacterium, Phascolarctobacterium, Buttiauxella, Bifidobacterium,Collinsella, Prevotella and Bacteroides genera. The alpha indexes showed a decrease in microbial diversity after 6 h of colonic fermentation. The coordinates analysis indicated any differences between irradiated and non-irradiated bar. The metabolic prediction demonstrated that MangoB and UVMangoB increase the microbiota carbohydrate metabolism pathway. This study suggests that IF of mango-based bar induced beneficial changes on microbial ecology and metabolic pathway that could be promissory to prevention or treatment of metabolic dysbiosis. However, in vivo interventions are necessary to confirm the interactions between microbiota modulating and intestinal beneficial effects.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tubos de membrana de celulosa para diálisis, avg. flat width 33 mm (1.3 in.)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Pancreatina from porcine pancreas, 4 × USP specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Aminoglucosidasa solution from Aspergillus niger, for use in Total Dietary Fiber Assay, TDF-100A
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
D-(+)-Rafinosa pentahydrate, ≥98% (HPLC), BioXtra
En este momento no podemos mostrarle ni los precios ni la disponibilidad