MilliporeSigma
[A man with a bump on his forehead after a fall].
Nederlands tijdschrift voor geneeskunde (2013-02-22)
Elisabeth G Klompenhouwer, Irene Thomassen, Joep A W Teijink
RESUMEN
An 84-year-old male patient presented with a swelling on his forehead which had developed gradually over a period of three weeks after a fall. Ultrasound examination revealed a complex fluid collection with a yin-yang flow pattern (revealed by colour Doppler imaging), which was suggestive of a pseudoaneurysm of the frontal branch of the superficial temporal artery. This was successfully treated with an injection of thrombin. traumatic pseudoaneurysm of the superficial temporal artery.