MilliporeSigma
  • Oxidative DNA damage from potassium bromate exposure in Long-Evans rats is not enhanced by a mixture of drinking water disinfection by-products.
Oxidative DNA damage from potassium bromate exposure in Long-Evans rats is not enhanced by a mixture of drinking water disinfection by-products.
Chemico-biological interactions (2005-04-21)
Kevin S McDorman, Brian F Pachkowski, Jun Nakamura, Douglas C Wolf, James A Swenberg
RESUMEN
Public drinking water treated with chemical disinfectants contains a complex mixture of disinfection by-products (DBPs) for which the relative toxicity of the mixtures needs to be characterized to accurately assess risk. Potassium bromate (KBrO(3)) is a by-product from ozonation of high-bromide surface water for production of drinking water and is a rodent carcinogen that produces thyroid, mesothelial, and renal tumors. The proposed mechanism of KBrO(3) renal carcinogenesis involves the formation of 8-oxoguanine (8-oxoG), a promutagenic base lesion in DNA typically removed through base excision repair (BER). In this study, male Long-Evans rats were exposed via drinking water to carcinogenic concentrations of KBrO(3) (0.4 g/L), 3-chloro-4-(dichloromethyl)-5-hydroxy-2(5H)-furanone (0.07 g/L), chloroform (1.8 g/L), bromodichloromethane (0.7 g/L), or a mixture of all these chemicals at the same concentrations for 3 weeks. Half of one kidney was processed for microscopic examination, and the remaining kidney was frozen for isolation of genomic DNA. Levels of 8-oxoG were measured using HPLC with electrochemical detection in DNA samples incubated with formamidopyrimidine-DNA glycosylase. Aldehydic lesions (e.g. abasic sites) in DNA samples were quantitated using an aldehyde-reactive probe slot-blot assay. Treatment with KBrO(3) produced a measurable increase of 8-oxoG in the kidney, and this effect was greater than that produced by treatment with the DBP mixture. No other single chemical treatment caused measurable increases of 8-oxoG. The mixture effect on the amount of 8-oxoG observed in this study suggests an interaction between chemicals that reduced the generation of oxidative DNA damage. No increases in abasic sites were observed with treatment, but a decrease was apparent in the rats treated with the DBP mixture. These data are consistent with previous studies where chronic exposure to this chemical mixture in drinking water resulted in a less than additive carcinogenic response in Tsc2 mutant Long-Evans rats.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Bromodichloromethane, ≥97%
En este momento no podemos mostrarle ni los precios ni la disponibilidad