MilliporeSigma
  • Structurally unique inhibitors of human mitogen-activated protein kinase phosphatase-1 identified in a pyrrole carboxamide library.
Structurally unique inhibitors of human mitogen-activated protein kinase phosphatase-1 identified in a pyrrole carboxamide library.
The Journal of pharmacology and experimental therapeutics (2007-06-01)
John S Lazo, John J Skoko, Stefan Werner, Branko Mitasev, Ahmet Bakan, Fumito Koizumi, Archibong Yellow-Duke, Ivet Bahar, Kay M Brummond
RESUMEN
Mitogen-activated protein kinase phosphatase 1 (MKP-1) is a tyrosine phosphatase superfamily member that dephosphorylates and inactivates cardinal mitogen-activated protein kinase (MAPK) substrates, such as p38, c-Jun NH(2)-terminal kinase, and extracellular signal-regulated kinase. Although these MAPK substrates regulate many essential cellular processes associated with human diseases, few pharmacological inhibitors have been described. The lack of readily available selective MKP-1 inhibitors has severely limited interrogation of its biological role and was one rationale for using a recently described tricyclic pyrrole-2-carboxamide library in our screening efforts. In this report we demonstrate the pharmacological richness of the pyrrole carboxamide library by the finding that 10 of 172 members inhibited human MKP-1. Two of the pyrrole carboxamides, PSI2106 and MDF2085, were especially notable in vitro inhibitors of recombinant human MKP-1 enzyme activity with IC(50) values of 8.0 +/- 0.9 and 8.3 +/- 0.8 microM, respectively. Both showed some selectivity for MKP-1 over the closely related phosphatases MKP-3, Cdc25B, VHR, and PTP1B. Computational examination of the surface properties near the catalytic site revealed that the phosphatases studied differ significantly in their electrostatic potential at the substrate binding site. The compounds inhibited MKP-1 reversibly but displayed mixed kinetics. Phosphatase inhibition was retained in the presence of physiologically relevant concentrations of glutathione. Molecular docking studies suggested that PSI2106 may interact with His(229) and Phe(299) on MKP-1. These results reveal the power of using a small focused library for identifying pharmacological probes.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Pyrrole-2-carboxylic acid, 99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad