MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
[Effect of growth factors and some microelements on biosurfactant synthesis of Acinetobacter calcoaceticus IMV B-7241].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2014-02-01)
T P Pirog, T A Shevchuk, O Iu Mashchenko, S A Parfeniuk, G A Iutinskaia
RESUMEN
The effect of yeast autolysate and microelements on synthesis of surface-active substances (SAS, biosurfactants) was investigated under cultivation of Acinetobacter calcoaceticus IMV B-7241 on various carbon substrates (n-hexadecane, ethanol, glycerol). The authors have shown a possibility to substitute the yeast autolysate and microelement mixture in the composition of ethanol- and n-hexadecane-containing media by copper sulfate (0.16 micromol/l) and iron sulfate (3.6 micromol/l), and in the medium with glycerol by 0.21 mmol/l of KCl, 38 micromol/l of zinc sulfate and 0.16 micromol/l of copper sulfate. Under such conditions of cultivation of the strain IMV B-7241 the SAS concentration exceeded that on the initial media, which contained the yeast autolysate and microelements, 1.2-1.6 times. The authors have also established the activating effect of low (0.01 mM) concentrations of Fe2+ on activity of the enzymes of biosynthesis of surface-active amino- (NADP-dependent glutamate dehydrogenase) and glycolipids (phosphoenolpyruvate(PhEP)-synthetase, PhEP-carboxykinase), as well as of anaplerotic reaction(PhEP-carboxylase). A necessity to introduce zinc cations into glycerol-containing medium is determined by their stimulating effect on activity of 4-dinitroso-N,N-dimethylaniline-dependent alcohol dehydrogenase--one of the enzymes of this substrate catabolism in A. calcoaceticus IMV B-7241.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Alcohol etílico puro 200, Molecular Biology
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Glicerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Alcohol etílico puro, 200 proof
Sigma-Aldrich
Glicerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glicerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sulfato de cinc heptahidratado, ACS reagent, 99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) sulfate, ReagentPlus®, ≥99%
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Glicerol, ≥99.5%
Sigma-Aldrich
Sulfato de cinc heptahidratado, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Glicerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Sigma-Aldrich
Reagent Alcohol, denatured, suitable for HPLC
Sigma-Aldrich
Glicerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glicerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glicerol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Sulfato de cinc heptahidratado, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Glicerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Sulfato de cinc heptahidratado, BioReagent, suitable for cell culture
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Sigma-Aldrich
Copper(II) sulfate solution, 4 % (w/v) (prepared from copper (II) sulfate pentahydrate)
Supelco
Etanol, Pharmaceutical Secondary Standard; Certified Reference Material