MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
[Diabetes mellitus and osteoporosis. Role for vitamin D in glucose and energy metabolism].
Clinical calcium (2012-08-31)
Daisuke Inoue
RESUMEN
Vitamin D insufficiency has been linked to obesity, type 2 diabetes, metabolic syndrome and dyslipidemia. The data, however, is mostly derived from observational studies and only indicates correlation. Evidence from randomized controlled trials, which are necessary to prove causal relationship between vitamin D insufficiency and those metabolic abnormalities, is scarce. Thus, at this point, although substantial circumstantial evidence supports potential roles of vitamin D in glucose and energy metabolism, clinical decisions on vitamin D treatment should not be based on such non-skeletal effects including protection from cardiovascular events, according to the recent clinical guidelines.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
D-(+)-Glucosa, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucosa, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucosa solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dextrosa, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glucosa solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucosa, ≥99.5% (GC), BioXtra
Supelco
D-(+)-Glucosa, Pharmaceutical Secondary Standard; Certified Reference Material
USP
D-(+)-Glucosa, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Glucosa, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucosa, ACS reagent
Supelco
D-(+)-Glucosa, analytical standard
Sigma-Aldrich
D-(+)-Glucosa, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucosa, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucosa, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucosa, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucosa solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-(+)-Glucosa solution, 1000 mg/dL