MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
SERPINB2 down-regulation contributes to chemoresistance in head and neck cancer.
Molecular carcinogenesis (2013-05-11)
Zhiquan Huang, Haigang Li, Qi Huang, Dan Chen, Jingjing Han, Lili Wang, Chanbin Pan, Weiliang Chen, Michael G House, Kenneth P Nephew, Zhongmin Guo
RESUMEN
Resistance to cisplatin-based chemotherapy is responsible for the majority of deaths from head and neck squamous cell carcinoma (HNSCC). In this study, using genome-wide gene expression analysis to investigate potential molecular mediators of HNSCC chemoresistance, we identified SERPINB2, a known inhibitor of extracellular serine proteinase urokinase-type plasminogen activator (uPA), as an important candidate. Whereas SERPINB2 is known to function as a suppressor of uPA molecular cascades, many of which play important roles in tumor invasion and metastasis, a role for SERPINB2 in cancer drug resistance has not been examined. By using quantitative real-time PCR and Western blot analysis, we determined that SERPINB2 mRNA and protein levels correlated with chemoresistance in HNSCC cell lines, and significantly lower SERPINB2 expression levels were observed in two cisplatin resistant HNSCC subclones compared to their isogenic drug-sensitive parental lines. Immunohistochemical analysis of HNSCC tumor tissues from patients treated with neoadjuvant cisplatin-based chemotherapy (n = 67 cases) revealed a significant association between SERPINB2 protein levels, tumor differentiation and patient relapse. Moreover, SERPINB2 down-regulation was a strong predictor of reduced overall survival in patients with HNSCC who received cisplatin-based chemotherapy (P = 0.001, log rank test). Studies using either siRNA-mediated down-regulation or forced over-expression of SERPINB2 in HNSCC cell lines confirmed a functional role for SERPINB2 in drug resistance. The findings were further supported using chemical inhibitors of STAT3 activity (a downstream effecter of uPAR signaling pathway), showing that STAT3 suppression altered HNSCC cell line cisplatin sensitivity. This is the first report on a role for SERPINB2 in acquired resistance to cisplatin in patients with HNSCC.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dimetilsulfóxido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimetilsulfóxido, Molecular Biology
Sigma-Aldrich
Dimetilsulfóxido, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Peróxido de hidrógeno solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Dimetilsulfóxido, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, anhydrous, ≥99.9%
Sigma-Aldrich
Peróxido de hidrógeno solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Peróxido de hidrógeno solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Dimetilsulfóxido, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Peróxido de hidrógeno solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Millipore
Peróxido de hidrógeno solution, 3%, suitable for microbiology
Supelco
Peróxido de hidrógeno solution, ≥30%, for trace analysis
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Peróxido de hidrógeno solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Dimetilsulfóxido, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Peróxido de hidrógeno solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Peróxido de hidrógeno solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Dimetilsulfóxido, PCR Reagent
Sigma-Aldrich
Peróxido de hidrógeno solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Peróxido de hidrógeno solution, 34.5-36.5%
Supelco
Peróxido de hidrógeno solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., dried, ≤0.02% water
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis