MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Toxicoproteomic analysis of pulmonary carbon nanotube exposure using LC-MS/MS.
Toxicology (2015-01-20)
Gina M Hilton, Alexia J Taylor, Christina D McClure, Gregory N Parsons, James C Bonner, Michael S Bereman
RESUMEN
Toxicoproteomics is a developing field that utilizes global proteomic methodologies to investigate the physiological response as a result of adverse toxicant exposure. The aim of this study was to compare the protein secretion profile in lung bronchoalveolar lavage fluid (BALF) from mice exposed to non-functionalized multi-walled carbon nanotubes (U-MWCNTs) or MWCNTs functionalized by nanoscale Al2O3 coatings (A-MWCNT) formed using atomic layer deposition (ALD). Proteins were identified using liquid chromatography tandem mass spectrometry (LC-MS/MS), and quantified using a combination of two label-free proteomic methods: spectral counting and MS1 peak area analysis. On average 465 protein groups were identified per sample and proteins were first screened using spectral counting and the Fisher's exact test to determine differentially regulated species. Significant proteins by Fisher's exact test (p<0.05) were then verified by integrating the intensity under the extracted ion chromatogram from a single unique peptide for each protein across all runs. A two sample t-test based on integrated peak intensities discovered differences in 27 proteins for control versus U-MWCNT, 13 proteins for control versus A-MWCNT, and 2 proteins for U-MWCNT versus A-MWCNT. Finally, an in-vitro binding experiment was performed yielding 4 common proteins statistically different (p<0.05) for both the in-vitro and in-vivo study. Several of the proteins found to be significantly different between exposed and control groups are known to play a key role in inflammatory and immune response. A comparison between the in-vitro and in-vivo CNT exposure emphasized a true biological response to CNT exposure.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ácido fórmico, reagent grade, ≥95%
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrilo, ACS reagent, ≥99.5%
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrilo, anhydrous, 99.8%
Sigma-Aldrich
Ácido fórmico, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ácido fórmico, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥88%
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrilo, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Metanol, Absolute - Acetone free
USP
Metanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Metanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Metanol, BioReagent, ≥99.93%
Supelco
Metanol, analytical standard
Supelco
Metanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrilo, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrilo, electronic grade, 99.999% trace metals basis