MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Enhanced retention and anti-tumor efficacy of liposomes by changing their cellular uptake and pharmacokinetics behavior.
Biomaterials (2014-12-20)
Yan Li, Ruiyuan Liu, Jun Yang, Yuanjie Shi, Guanghui Ma, Zhenzhong Zhang, Xin Zhang
RESUMEN
Although PEGylated liposome-based drug delivery systems hold great promising applications for cancer therapy due to their prolonged blood circulation time, PEGylation significantly reduces their cellular uptake, which markedly impairs the in vivo tumor retention and antitumor efficiency of drug-loaded liposomes. Most importantly, it has been proved that repeated injections of PEGylated liposomes with cell cycle specific drug such as topotecan (TPT) in the same animal at certain time intervals will induce "accelerated blood clearance" (ABC) phenomenon, which decreases the tumor accumulation of drug-loaded liposomes and presents a tremendous challenge to the clinical use of liposome-based drug delivery systems. Herein, we developed a zwitterionic poly(carboxybetaine) (PCB) modified liposome-based drug delivery system. The presence of PCB could avoid protein adsorption and enhance the stability of liposomes as that for PEG. Quite different from the PEGylated liposomes, the pH-sensitive PCBylated liposomes were internalized into cells via endocytosis with excellent cellular uptake and drug release ability. Furthermore, the PCBylated liposomes would avoid ABC phenomenon, which promoted the tumor accumulation of drug-loaded liposomes in vivo. With higher tumor accumulation and cellular uptake, the PCBylated drug-loaded liposomes significantly inhibited tumor growth and provided a promising approach for cancer therapy.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Alcohol etílico puro 200, Molecular Biology
Sigma-Aldrich
Ácido trifluoroacético, ReagentPlus®, 99%
Sigma-Aldrich
Ácido trifluoroacético, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ácido acético, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Alcohol etílico puro, 200 proof
Sigma-Aldrich
Ácido acético, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrilo, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrilo, anhydrous, 99.8%
Sigma-Aldrich
Ácido trifluoroacético, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Colesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ácido acético, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, ACS reagent, ≥98%
Sigma-Aldrich
Ácido acético solution, suitable for HPLC
Sigma-Aldrich
Ácido acético, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Ácido acético, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Doxorrubicina hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ácido trifluoroacético, ≥99%, for protein sequencing
Sigma-Aldrich
Colesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Acetonitrilo, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Ammonium phosphate monobasic, ReagentPlus®, ≥98.5%
Sigma-Aldrich
Doxorrubicina hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)