MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Exploring the conformational variability in the heme b propionic acid side chains through the effect of a biological probe: a study of the isolated ions.
Exploring the conformational variability in the heme b propionic acid side chains through the effect of a biological probe: a study of the isolated ions.
The journal of physical chemistry. B (2015-01-13)
Alberto De Petris, Barbara Chiavarino, Maria Elisa Crestoni, Cecilia Coletti, Nazzareno Re, Simonetta Fornarini
RESUMEN
The iron(III) protoporphyrin IX complex with imidazole, a biologically relevant ligand, occupying an axial position, has been studied by infrared multiple photon dissociation (IRMPD) spectroscopy. The complex has been delivered in gas-phase by electrospray ionization (ESI), mass selected in an ion trap, and assayed by IRMPD spectroscopy in two complementary frequency regions. The fingerprint range (900-1900 cm(-1)) has been scanned using the Orsay free-electron laser beamline (CLIO), while the X-H (X = C,N,O) stretching region (3000-3600 cm(-1)) has been inspected using a tabletop IR optical parametric oscillator/amplifier (OPO/OPA) laser source. DFT calculations have been performed to obtain a comprehensive pattern of the various potential conformers yielding optimized geometries, relative thermodynamic parameters, and respective IR spectra. The comparison between the IR spectra for representative conformers and the experimental IRMPD features suggests the coexistence of two families of conformers involving different degrees of folding and hydrogen bonding between the two propionic acid functionalities on the periphery of the protoporphyrin IX macrocycle in a ratio depending on environmental conditions such as ESI solvent and temperature. The observed conformational variability of the porphyrin substituents in the naked heme-imidazole complex is consistent with the fine-tuning of the reactivity properties of this important prosthetic group by the specific surroundings in the protein core.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Diclorometano, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Diclorometano, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrilo, ACS reagent, ≥99.5%
Sigma-Aldrich
Diclorometano, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrilo, anhydrous, 99.8%
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ftaldialdehído, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Diclorometano, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrilo, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Metanol, Absolute - Acetone free
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol