MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Evaluation of synergistic effect of biodegradable polymeric nanoparticles and aluminum based adjuvant for improving vaccine efficacy.
Evaluation of synergistic effect of biodegradable polymeric nanoparticles and aluminum based adjuvant for improving vaccine efficacy.
International journal of pharmaceutics (2014-06-19)
Vivek Bansal, Manoj Kumar, Manu Dalela, H G Brahmne, Harpal Singh
RESUMEN
Aluminum based adjuvants have been used widely to induce long lasting protective immunity through vaccination. But reported incidences of toxicity and side effects of aluminum have raised concerns regarding their safety in childhood vaccines. The present study demonstrates the synergistic effect of admixture of polylactic acid-polyethylene glycol (PLA-PEG) based biodegradable nanoparticles (NPs) and aluminum phosphate as a potential adjuvant system using tetanus toxoid (TT) as a model antigen. The immunological activity of the admixture formulation was maintained up to 180 days of storage at 5 °C±3 °C. Percent adsorption/encapsulation of tetanus toxoid increased to nearly 90% in admixture formulation as compared to 55% in conventional vaccine. Admixture preparation (PLA-PEG-Al 0.2 mg-TT and PLA-Al 0.2 mg-TT) showed 80% and 50% survival respectively, even at 180 days as compared to 30% survival observed in the conventional tetanus vaccine. The present study established the feasibility to formulate a dosage form with improved efficacy and reduced aluminum concentration for vaccination.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Diclorometano, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Diclorometano, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrilo, ACS reagent, ≥99.5%
Sigma-Aldrich
Diclorometano, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrilo, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Diclorometano, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrilo, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Diclorometano, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Supelco
Diclorometano, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrilo, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrilo, electronic grade, 99.999% trace metals basis
Supelco
Diclorometano, analytical standard
Sigma-Aldrich
Acetonitrilo solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrilo, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitrilo, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrilo solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Acetonitrilo, analytical standard
Supelco
Acetonitrilo, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrilo, ReagentPlus®, 99%
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Diclorometano, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)