MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Enterobacillus tribolii gen. nov., sp. nov., a novel member of the family Enterobacteriaceae, isolated from the gut of a red flour beetle, Tribolium castaneum.
Enterobacillus tribolii gen. nov., sp. nov., a novel member of the family Enterobacteriaceae, isolated from the gut of a red flour beetle, Tribolium castaneum.
Antonie van Leeuwenhoek (2015-02-27)
Vikas S Patil, Rahul C Salunkhe, Ravindra H Patil, C Husseneder, Yogesh S Shouche, V Venkata Ramana
RESUMEN
Two novel Gram-stain negative facultative anaerobic, motile, rod-shaped bacterial strains IG-V01(T) and IG-V01b were isolated from the gut of red flour beetles, Tribolium castaneum. The 16S rRNA gene sequences of strains IG-V01(T) and IG-V01b were found to have their highest sequence similarity (96.5% and 96.4%) with Serratia nematodiphila DZ0503SBS1(T) (Enterobacteriaceae family) respectively. Strains IG-V01(T) and IG-V01b share 100% 16S rRNA gene sequence similarity and exhibit very similar phenotypic characteristics. In addition, they show 89.7% genomic relatedness (DNA-DNA hybridisation). Major fatty acids were identified to be C(16:0) (38.3%), C(17:0) cyclo (19.5-20%) and C(14:0) (11.2-11.3%). Cells contain phosphatidylethanolamine and diphosphatidylglycerol as predominant polar lipids. Genomic DNA G+C content (mol%) was determined to be 51.5-51.7. A polyphasic approach employing the study of morphological, physiological, chemotaxonomic, genomic and phylogenetic analysis revealed that the two newly isolated strains cannot be placed in any of the existing genera of the family Enterobacteriaceae. Therefore, it is proposed that strains IG-V01(T) and IG-V01b belong to a novel genus within the family Enterobacteriaceae, and represent a new species Enterobacillus tribolii gen. nov., sp. nov., with the type strain =IG-V01(T) = KCTC 42159(T) = MCC 2532(T).
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Alcohol etílico puro 200, Molecular Biology
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ácido acético, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Alcohol etílico puro, 200 proof
Sigma-Aldrich
Ácido acético, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Cloroformo, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Ácido fórmico, reagent grade, ≥95%
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Cloroformo, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrilo, ACS reagent, ≥99.5%
Sigma-Aldrich
Cloroformo, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Cloroformo, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrilo, anhydrous, 99.8%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ácido fórmico, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ácido acético, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Ácido acético solution, suitable for HPLC
Sigma-Aldrich
Ácido acético, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Ácido acético, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Ácido fórmico, puriss., meets analytical specifications of DAC, FCC, 98.0-100%