MilliporeSigma
Characterization of cyclic peptides containing disulfide bonds.
Journal of pharmaceutical and biomedical analysis (2015-03-18)
Mindy Johnson, Mingtao Liu, Elaine Struble, Kanthi Hettiarachchi
RESUMEN
Unlike linear peptides, analysis of cyclic peptides containing disulfide bonds is not straightforward and demands indirect methods to achieve a rigorous proof of structure. Three peptides that belong to this category, p-Cl-Phe-DPDPE, DPDPE, and CTOP, were analyzed and the results are presented in this paper. The great potential of two dimensional NMR and ESI tandem mass spectrometry was harnessed during the course of peptide characterizations. A new RP-HPLC method for the analysis of trifluoroacetic acid is also presented. It is robust, simple, and efficient compared to the currently available methods.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ácido trifluoroacético, ReagentPlus®, 99%
Sigma-Aldrich
Ácido trifluoroacético, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Dimetil sulfóxido-d6, 99.9 atom % D
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrilo, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimetil sulfóxido-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Acetonitrilo, anhydrous, 99.8%
Sigma-Aldrich
Ácido trifluoroacético, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
DL-Ditiotreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
DL-Ditiotreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Dimetil sulfóxido-d6, 99.5 atom % D
Sigma-Aldrich
Ácido trifluoroacético, ≥99%, for protein sequencing
Sigma-Aldrich
DL-Ditiotreitol, BioUltra, Molecular Biology, ≥99.5% (RT)
Sigma-Aldrich
DL-Ditiotreitol, ≥99.0% (RT)
Sigma-Aldrich
Acetonitrilo, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
DL-Ditiotreitol, suitable for electrophoresis, ≥99%
Sigma-Aldrich
DL-Ditiotreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Dimetil sulfóxido-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Acetonitrilo, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrilo, electronic grade, 99.999% trace metals basis
Supelco
Ácido trifluoroacético, analytical standard
Sigma-Aldrich
Dimetil sulfóxido-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Acetonitrilo solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Dimetil sulfóxido-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Acetonitrilo, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitrilo, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrilo solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC