MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A possible antineoplastic potential of selective, irreversible proteasome inhibitor, carfilzomib on chemically induced hepatocarcinogenesis in rats.
A possible antineoplastic potential of selective, irreversible proteasome inhibitor, carfilzomib on chemically induced hepatocarcinogenesis in rats.
Journal of biochemical and molecular toxicology (2014-05-28)
Mahmoud A Mansour, Mohammed A Aljoufi, Khaled Al-Hosaini, Ammar C Al-Rikabi, Mahmoud N Nagi
RESUMEN
The antineoplastic effect of carfilzomib (CFZ) against chemically induced hepatocarcinogenesis was studied. A total of 60 male Wistar albino rats were divided into six groups with 10 animals in each group. Rats in group 1 (control group) were given dimethylsulphoxide (DMSO) (0.4 mL/kg i.p) twice a week for 3 weeks from week 8 to week 10. Animals in groups 2 and 3 were given CFZ (2 and 4 mg/kg i.p) twice a week from week 8 to week 10, respectively. Rats in group 4 were given diethylnitrosamine (DENA) at a dose of 0.01% in drinking water for 10 weeks and received a DMSO (0.4 mL/kg i.p) twice a week from week 8 to week 10. Animals in groups 5 and 6 were given DENA at a dose of 0.01% in drinking water for 10 weeks and treated with CFZ (2 and 4 mg/kg i.p) twice a week from week 8 to week 10, respectively. CFZ succeeded in suppressing the elevated serum tumor marker α-fetoprotein and carcinoembryonic antigen. The antineoplastic effect of CFZ was also accompanied by normalization of elevated hepatic tissue growth factors, matrix metalloproteinase-2 and tissue inhibitor of metalloproteinase-1, and augmentation of hepatic endostatin and metallothionein. A histopathological examination of liver samples treated with CFZ after DENA intoxication correlated with the biochemical observation. Treatment with CFZ confers an antineoplastic activity against chemically induced hepatocarcinogenesis. These findings suggest that CFZ plays a pivotal role in the treatment of hepatocarcinogenesis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dimetilsulfóxido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimetilsulfóxido, Molecular Biology
Sigma-Aldrich
Dimetilsulfóxido, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimetilsulfóxido, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, anhydrous, ≥99.9%
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimetilsulfóxido, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimetilsulfóxido, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimetilsulfóxido, PCR Reagent
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., dried, ≤0.02% water
Supelco
Dimetilsulfóxido, analytical standard
Supelco
Dimetilsulfóxido, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimetilsulfóxido, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, JIS special grade, ≥99.0%
Dimetilsulfóxido, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Human AFP / Alpha-fetoprotein ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Dimetilsulfóxido, Vetec, reagent grade, 99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimetilsulfóxido, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimetilsulfóxido, suitable for HPLC
Sigma-Aldrich
Dimetilsulfóxido, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimetilsulfóxido, SAJ first grade, ≥99.0%