MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Dual-cargo selectively controlled release based on a pH-responsive mesoporous silica system.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-12-11)
Wanyuan Gui, Wenqian Wang, Xiangyu Jiao, Lifeng Chen, Yongqiang Wen, Xueji Zhang
RESUMEN
A pH-controlled delivery system based on mesoporous silica nanoparticles (MSNs) was constructed for dual-cargo selective release. To achieve a better controlled-release effect, a modified sol-gel method was employed to obtain MSNs with tunable particle and pore sizes. The systems selectively released different kinds of cargo when stimulated by different pH values. At the lower pH value (pH 2.0) only one kind of cargo was released from the MSNs, whereas at a higher pH value (pH 7.0) only the other kind of cargo was released from the MSNs. The multi-cargo delivery system has brought the concept of selective release to new advances in the field of functional nanodevices and allows more accurate and controllable delivery of specific cargoes, which is expected to have promising applications in nanomedicine.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Alcohol etílico puro 200, Molecular Biology
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Alcohol etílico puro, 200 proof
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrilo, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrilo, anhydrous, 99.8%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ortosilicato de tetraetilo, reagent grade, 98%
Sigma-Aldrich
Borohidruro de sodio, powder, ≥98.0%
Sigma-Aldrich
Bromuro de hexadeciltrimetilamonio, ≥98%
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Bromuro de hexadeciltrimetilamonio, Molecular Biology, ≥99%
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Borohidruro de sodio, ReagentPlus®, 99%
Sigma-Aldrich
Ortosilicato de tetraetilo, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrilo, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Bromuro de hexadeciltrimetilamonio, BioXtra, ≥99%
Sigma-Aldrich
Borohidruro de sodio, granular, 99.99% trace metals basis
Supelco
Etanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Alcohol etílico puro, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ortosilicato de tetraetilo, 99.999% trace metals basis
Sigma-Aldrich
Borohidruro de sodio, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrilo, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrilo, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrilo solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Bromuro de hexadeciltrimetilamonio, BioUltra, Molecular Biology, ≥99.0% (AT)