Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41106514
eCl@ss:
32040503
Biological source:
human blood
Technique(s):
cell culture | mammalian: suitable
Ethnicity:
UK Caucasian
Gender:
female
Growth mode:
Suspension
Morphology:
Lymphoblastoid
Product Name
C2093, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
biological source
human blood
manufacturer/tradename
ECACC
packaging
tube of 5 μg 950405166-DNA-5UG, pkg of vial of cells 950405166-1VL
HRC Panel Number
4
ethnicity
UK Caucasian
gender
female
growth mode
Suspension
karyotype
Not Specified
morphology
Lymphoblastoid
technique(s)
cell culture | mammalian: suitable
Application
As part of the Human Random Control collection this cell line can be used as a renewable source of genomic DNA reference material for a wide range of genetic research applications.
Biochem/physiol Actions
A B-lymphoblastoid cell line derived from a randomly selected Caucasian blood donor whose parents and grandparents were born in the UK or Ireland. The cell line was generated by Epstein-Barr Virus (EBV) transformation of peripheral blood lymphocytes.
This cell line is part of the European Collection of Cell Cultures (ECACC) Human Random Control (HRC) collection. The HRC collection consists of more than 700 B-lymphoblastoid cell lines derived from randomly selected Caucasian blood donors whose parents and grandparents were born in the UK or Ireland. The HRC collection can be used as a renewable control population for genetic research.
This cell line is part of the European Collection of Cell Cultures (ECACC) Human Random Control (HRC) collection. The HRC collection consists of more than 700 B-lymphoblastoid cell lines derived from randomly selected Caucasian blood donors whose parents and grandparents were born in the UK or Ireland. The HRC collection can be used as a renewable control population for genetic research.
UK Caucasian Female
Preparation Note
RPMI 1640 (R0883) + 2 mM L-Glutamine (G7513) + 20% FBS / FCS (F2442). Once a growing culture is established the FBS concentration can be reduced to 10%.
Maintain cultures between 3 x 105 - 2 x 106 cells/ml; 5% CO2, 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.