Saltar al contenido
Merck

Saltar a

DWK356830

WHEATON® CELSTIR MAGNA FLEX Spinner Flask

capacity 125 mL, bulb spinner, double sidearm, with cap, clear, pkg of 1 ea

Sinónimos:

356830, CELSTIR, culture flask, flask, spinner flask, stir flask, suspension culture flask, vessel

Iniciar sesión para ver los precios por organización y contrato.

Seleccione un Tamaño

Cambiar Vistas
A ustedes/SKUDisponibilidadPrecio

Acerca de este artículo

UNSPSC Code:
41121804
NACRES:
NB.43
Capacity:
125 mL
Material:
borosilicate glass 3.3 flask, clear
Packaging:
pkg of 1 ea
Feature:
autoclavable, with cap, bulb spinner, double sidearm
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarle


material

borosilicate glass 3.3 flask, clear

joint

joint threaded center 51-400, joint threaded side arm 33-430

feature

autoclavable, with cap, bulb spinner, double sidearm

packaging

pkg of 1 ea

manufacturer/tradename

DWK Life Sciences WHEATON® 356830

capacity

125 mL

suitability

suitable for (Suspension cell culture)

General description

The CELSTIR Spinner Flask is constructed from borosilicate glass 3.3 and is specifically designed for suspension cell culture applications with a magnetic stirrer. It features an adjustable paddle blade impeller for enhanced mixing, along with Magna-Flex™ impellers that are bulb-shaped to provide gentle stirring while maintaining a sterile environment, as the stirrer shaft does not protrude through the cap. The flask has a 1:1 headspace ratio for optimal gas exchange, and its bottom dimple design improves circulation and minimizes cell accumulation in the center. Additionally, the entire unit can be autoclaved at 121°C for effective sterilization.

Application

cell culture, suspension cell culture

Features and Benefits

  • Manufactured from borosilicate glass 3.3
  • The CELSTIR Spinner Flask features an adjustable paddle blade impeller for better mixing.
  • For suspension cell culture with a magnetic stirrer
  • Adjustable paddle impeller and sterile design as stirrer shaft does not protrude through cap
  • 1:1 headspace ratio for ideal gas exchange
  • Bottom dimple improves circulation and reduces the accumulation of cells in the center of the flask (dimple on 125mL and larger flasks)
  • The entire unit may be autoclaved at 121°C
  • Magna-Flex impellers are bulb shaped providing a gentle stirring

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.


Still not finding the right product?




Elija entre una de las versiones más recientes:

Certificados de análisis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documentos section.

Si necesita más asistencia, póngase en contacto con Atención al cliente

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos