Skip to Content
MilliporeSigma

MTCR2392

MTC™ Bio DoubleDecker™ Cardboard PCR Freezer Box

to hold, 392 x 0.1 mL or 0.2 mL PCR tubes, strips or plates, pack of 5 ea

Synonym(s):

cryovial storage, MTC Bio R2392, cardboard freezer box, cryo box, freezer storage

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41123300
Manufacturer/tradename:
MTC™ Bio R2392
Feature:
lid hinged, to hold 392 x 0.1 mL or 0.2 mL PCR tubes, strips or plates
Material:
cardboard
Packaging:
pack of 5 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

cardboard

description

format 14 x 14

feature

lid hinged, to hold 392 x 0.1 mL or 0.2 mL PCR tubes, strips or plates

packaging

pack of 5 ea

manufacturer/tradename

MTC™ Bio R2392

well size

196 , on each level

General description

This unique freezer box has been designed to maximize storage capacity for PCR tubes. With 196 wells on each level, it can accommodate 392 individual 0.1mL or 0.2mL tubes. For strip tubes, the box handles 40 strips of 8 or 28 strips of 12. The top level is a cardboard tray with two finger grips that allow easy access to the lower level. The two level system doubles the capacity while keeping the total box height under 2 inches (51mm) and assuring a good fit in standard height stainless steel racks.

Application

PCR

Features and Benefits

  • 196 wells on each level
  • Can hold 392 individual 0.1mL or 0.2mL tubes.
  • Can hold 40 strips of 8 or 28 strips of 12
  • Doubles capacity
  • Height under 2 in (51 mm)

Legal Information

DoubleDecker is a trademark of MTC Bio
MTC is a trademark of MTC Bio




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library