MilliporeSigma
Primary Cell Culture
The image appears to be a microscopic view showing various cellular structures. It features cells with blue nuclei, green cytoskeleton fibers, and red mitochondria.
Primary cells are derived directly from living tissue, providing researchers with more meaningful models of human health and disease and more closely replicating the physiology of biological systems. They are increasingly useful in life science research, ADME/toxicity studies, and pharmaceutical development, and in diverse applications where cell lines do not replicate the target biological system or where immortalized cell lines have attributes that can confound results.
Our collection of primary cells comes with the promise of purity, low passage, rigorous characterization, and strict quality control. Harmonized media and reagents provide optimal performance, ensuring confidence in the utility of these cells and their supporting products in your application. We offer PromoCell® human primary cells (available in select geographies), primary blood and immune cells from BioIVT, and primary cells from Cell Applications, Inc. (CAI). See below for more information on our primary cell portfolios.
Products
Sort by Relevance
Showing 1-20 of 278
Change View
Showing 1-20 of 278
Compare
Product Number
Product Name
Product Description
Pricing
It shows a clear view of the heart, arteries, and veins in blue and red colors, which represent the pathways of oxygenated and deoxygenated blood, respectively.
PROMOCELL® Human Primary Cell Culture
PromoCell® products are available in North America, Canada, and select countries in Europe and Asia. 
Our primary cell collection includes cells and precisely formulated media from industry-leading manufacturer PromoCell®. We offer a broad range of validated, ethically-sourced human primary and stem cells, plus human blood cells—along with culture media optimized for expert culture of these phenotypes. 
  • Authenticated primary cells from dozens of human tissue sources, with media formulated for successful culture
  • Expertly-crafted primary cell culture protocols, including for tumor spheroid formation, culturing human cardiac myocytes, generating organotypic air-liquid interface (ALI) 3D skin cultures, and more
  • Advanced media and reagents for 3D culture from patient cancers
  • Ethically collected human primary cells with donor documentation, and validated for identity with definitive phenotypic markers
  • Primary cells from relevant human tissues for disease modeling in Diabetes Type I & II and Respiratory Diseases (COPD Type I–IV, Asthma), supporting translational research and therapeutic development
Unique formulations for groundbreaking cancer studies include the Primary Cancer Culture System, a complete solution for the selective culture of malignant cells extracted from primary tumors or patient xenografts.
BioIVT Elevating Science™ Logo
BioIVT peripheral blood mononuclear cells (PBMCs) and immune cell subsets 
In partnership with BioIVT, we offer an extensive collection of ready-to-use normal and disease state PBMCs as well as a variety of pre-isolated immune cell subsets with guaranteed purity, sterility, and yield. Our PMBCs and immune cells can be stored in the freezer until needed, with a demonstrated viability and functionality.
  • Wide range of cryopreserved PBMCs and immune cell subtypes including B cells, T cells, and NK cells for applications such as ADCC and T cell killing assays, drug toxicity assays, and more.
  • HLA typing available for many cell types
  • Request specific donor information to meet study-specific inclusion and exclusion donor criteria
Get answers to your questions about PMBCs and immune cells with our frequently asked questions technical article.
Click Here to Request Information about specific donor demographics or other criteria.
Cell Applications Inc. Primary Cells
Cell Applications Inc. has perfected the isolation, purification, subculture and growth of human and animal primary cells. Diverse primary cell options include cryopreserved formats, and cells prescreened for metabolic regulation markers and signaling pathways.
Diverse primary cell options include cryopreserved formats, and cells prescreened for metabolic regulation markers and signaling pathways.
Related Resources
Page 1 of 3