MilliporeSigma
Nucleosides and Nucleotides
Illustration of the location and structure of DNA double helix highlighting components that hold the gene specificity of every living being.
Nucleosides and nucleotides and their derivatives are biologically ubiquitous substances that participate in nearly all biochemical processes. They form the monomeric units of nucleic acids and thereby play central roles in both the expression and storage of genetic information.
Nucleosides are glycosylamines obtained by chemical or enzymatic decomposition of nucleic acids and contain two components: afive-carbon sugar (riboseor2’ deoxyribose) and anitrogen base. The nitrogenous bases are planar, aromatic, heterocyclic molecules. For the most part, are derivatives of purine or pyrimidine. The majorpurinecomponents of nucleic acids areadenine (A) andguanine (G) residues and the major pyrimidine residues arecytosine (C), Uracil (U) which mainly occurs in RNA), andthymine1.In double-helical DNAs and RNAs, the base compositions obey Chargaff’s rules: A=T(U) and G=C1.Examples of nucleosides include adenosine, guanosine, cytidine, uridine, and deoxythymidine. They function as signaling molecules and are precursors for nucleotides needed for DNA and RNA synthesis. Nucleosides also play a vital role in medicine and molecular biology and are used as antiviral or anticancer agents1.
Products
Sort by Relevance
Showing 1-20 of 268
Change View
Showing 1-20 of 268
Compare
Product Number
Product Name
Product Description
Pricing
As the building block of RNA and DNA, nucleotides are organic molecules composed of three components: a five-carbon sugar, a nitrogen base and, one or more phosphate groups. The nitrogen bases in the great majority of nucleotides are the purines (Adenine + Guanine) and the pyrimidines (Cytosine, either Thymine in DNA or Uracil in RNA). Examples of nucleotides include Adenosine monophosphate (AMP), Guanosine monophosphate (GMP), Cytidine monophosphate (CMP), Uridine monophosphate (UMP), and Deoxythymidine monophosphate (dTMP)1. Nucleotides are not only the basic unit of genetic material in all living things, but they also play a role in the energy, metabolism, and signaling in cells. In addition to their biological functions, nucleotides, and their derivatives play an important role in medical applications. By using the synthetic purine and pyrimidine analogs that contain halogens, thiols, or additional nitrogen atoms in the chemotherapy of cancer and AIDS, and immune suppressors during organ transplantation2.
Cell Biology and Drug Discovery
We offer a large selection of natural and synthetic nucleosides, nucleotides, oligonucleotides, and their analogs for a multitude of research, diagnostic and manufacturing applications in cell biology and drug discovery. We’re dedicated to supporting your scientific advancement with our innovative and reliable bioreagents available in mg to kg batch sizes to precisely meet your needs.
Breakthroughs feel closer than ever with our unparalleled portfolio:
  • Adenosine-derived nucleosides and nucleotides
  • Metabolic nucleotides
  • Cancer and proliferation biomarker nucleosides
  • Non-hydrolyzable nucleotide analogs
  • Coenzymes and essential cofactors
  • Cell culture, cell-arrest, cell inhibitor nucleosides
  • Protein purification nucleosides
  • Molecular & HPLC standards
References
1.
Donald V, Judith G. V. 2011. Biochemistry. 4th ed.. New Jersey: John Wiley & Sons.
2.
V.W. R. 2016. Nucleotides. Harper's Illustrated Biochemistry, 30e. McGraw Hill.