MilliporeSigma
Single-Use Assemblies
Mobius® 2D and 3D Bag Assemblies
Our Mobius® 2D and 3D assemblies provide efficient and scalable bioprocessing fluid management solutions, from media and buffer preparation to final fill applications. With a full spectrum of standard and custom solutions, at varying levels of certification, you get greater flexibility, mobility, and support for your single-use technologies. Our assemblies and solutions are designed to reduce operator error and process risks while improving efficiency.
  • Mobius® Standard 2D Assemblies, are sterilized and ready to use in a wide range of applications.
  • Mobius® Standard 3D Drum Assemblies, feature Pureflex™ films and are sterilized and ready to use for fluid storage or processing applications.
  • Mobius® Standard 3D Bin Assemblies are made with Ultimus® film or Pureflex™ flms, feature our EZ Fold technique and are designed for easy deployment for transport and storage.
  • Mobius® 2D Freeze Assemblies for frozen Liquid Transportation and Storage are made with our PureFlex™ film and Helium integrity tested to assure integrity at temperatures down to -80 °C.
Section Overview
Sort by Relevance
Showing 1-20 of 57
Change View
Showing 1-20 of 57
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 57 results
Ready-to-ship Single-use Assemblies Help Meet Your Challenging Timeline
Mobius® Essential Assemblies are standard, off-the-shelf, simple bioprocess bag and tubing assemblies. These plug-and-play assemblies are made of our proven PureFlex™ film and are available in both 2D bags (1–50 L), 3D bags (100–200 L), as well as bioprocess tubing assemblies. These standard assemblies are easily integrated into your current processes providing you with a straight path for scale-up to our Mobius® MyWay assemblies with PureFlex™ film.
  • Eliminate design and manufacturing lead time
  • 1 L - 200 L sizes that can easily scale up to premium assemblies under Mobius® MyWay program
Need help to find the right Mobius® Essential Assemblies for your processing needs? Fill out a short form to connect with our single-use specialist today.
Mobius® MyWay – Options to Suit Your Supply Needs
Mobius® MyWay is a comprehensive three-tiered program designed to promote a balance of single-use lead times with design flexibility, while enhancing supply security. Mobius® MyWay allows you to have the flexibility of ordering standard and configurable “fit for purpose” assemblies while still receiving the benefits of documentation support and extractable data at reduced lead times. Our unique approach reduces consumable management burden and cost, enhances supply transparency, and improves security.
MOBIUS® STOCK
Icon with a turquoise hexagonal chain design inside a shield-like shape.
    MOBIUS® SELECT
    Icon featuring interconnected geometric shapes including hexagons and triangles within a shield-like shape
      MOBIUS® CHOICE
      Icon with three upward arrows inside a shield-like shape
        Off-the-Shelf Standard Assemblies 
        Configure-to-Order Assemblies, Using a Library of Prequalified Components 
        Highly Customized Solutions
        Mobius® Assembly Certification & Release Testing
        Mobius® assemblies are available in several levels of certification. Certification of an assembly is based on the qualification of the components in the assembly, the level of leak testing performed during manufacturing, and the testing performed on the assembly lot after manufacturing. The certification level also impacts the shelf life and sterility claims that are made for the assembly.
        Helium Integrity Manufacturing Release Test for Increased Integrity Assurance
        Mobius® single-use assemblies* can also be tested with a Helium Integrity Manufacturing Release Test. Our Helium Integrity Test is validated to detect defects as small as 2 µm, and is designed to test the entire single-use assembly, including tubing and connection points. Incorporating the Helium Integrity Test into your integrity assurance strategy reduces the risk of leaks or microbial ingress in your manufacturing process.
        Mobius® assemblies for Helium Integrity Test is subject to the design space rules. Please find the details in the Helium Test Data Sheet.
        Certification LevelShelf LifeSterilityUSP <85> Endotoxins,
        USP <788> Sub-visible Particulates
        Leak TestingUSP <88> Biological Reactivity in vivo (Class VI),
        USP <661> Physicochemical Tests
        GOLD + HELIUM2 yearsSterilized using a validated gamma irradiation level of 25-40 kGy1 assembly per lot tested, post-gamma irradiation100% assembly helium integrity testedPost-gamma irradiated
        GOLD2 yearsSterilized using a validated gamma irradiation level of 25-40 kGy1 assembly per lot tested, post-gamma irradiation100% assembly leak testedPost-gamma irradiation
        SILVER2 yearsSterilized using a validated gamma irradiation level of 25-40 kGyQuarterly review of results, post-gamma irradiationIn-process testingPost-gamma irradiation
        BRONZENo claimExposed to gamma irradiation level of 25-40 kGyNo testing performedNo testing performedPre-gamma irradiation
        BASICNo claimNon-sterileNo testing performedIn-process testingPre-gamma irradiation
        Single-use Film Technology
        Our Ultimus® film was designed for superior robustness and durability in large-volume processing through a proprietary woven nylon structure. The fluid contact layer is animal origin-free, with low extractable, and supports healthy cell growth as it does not contain Irgafos® 168. 
        Our PureFlex™ films are used for the construction of Mobius® single-use 2D and 3D bag assemblies. Both PureFlex™ and PureFlex™ Plus films consist of a single fluid contact layer with a low extractable profile, chemically resistant materials, and zero animal-derived components, offering the flexibility and robustness you need throughout your single-use manufacturing processes.
        Have a question about Ultimus® film? Contact our technical experts. 
        Mobius® Single-use Assemblies are Supported by the Emprove® Program – The Smart Way To Master Compliance And Control
        Complementing our product portfolio, the Emprove® Program provides convenient access to reliable technical, regulatory and supply information in Emprove® Dossiers to support your risk assessment continuum. Our Advanced Qualification Dossier is customized with specific information for your single-use assembly.
        Bioprocess Risk Mitigation
        With a global network of sales development specialists and engineers we can support you with system demonstration, design and implementation, at-scale process simulation, and feasibility studies, as well as validation studies and documentation, supported by Millipore® validation services.
        Related Resources
        Page 1 of 3