MilliporeSigma
HomeMass SpectrometryAnalysis of Acylcarnitines in Dried Blood Spots (DBS) Samples by FIA-MS/MS
Analysis of Acylcarnitines in Dried Blood Spots (DBS) Samples by FIA-MS/MS
Arun Babu Kumar Ph.D.1, Sarah Aijaz Ph.D.1, Uma Sreenivasan Ph.D.1 MilliporeSigma 1 Round Rock, TX, USA
Workflow
Extraction
Sample extraction and derivatization
Sample extraction and derivatization
    Analysis
    FIA-MS/MS Analysis
    FIA-MS/MS Analysis
      Detection
      Detection & Quantification
      Detection & Quantification
        1. Overview: FIA-MS/MS for Acylcarnitines Analysis in Dried Blood Spots (DBS)
        In the 1960s, Robert Guthrie implemented the large-scale use of dried blood spots (DBS) for the screening of newborns for Phenylketonuria (PKU). Since then, the use of DBS in diagnosis has expanded to include a variety of metabolic disorders and other ailments. Tandem mass spectrometry (MS/MS) has developed to be the choice of technique to analyze the DBS samples due to its sensitivity and capacity to multiplex to quantify several analytes simultaneously. MS/MS is widely used in clinical labs for the analysis of acylcarnitine panel in DBS to diagnose several fatty acid oxidation disorders and organic acid disorders.1-3 Flow injection analysis-MS/MS (FIA-MS/MS) is widely used in Tier-1 testing for identification and quantitation of acylcarnitines by direct quantitation against suitable internal standards. 4,5 Figure 1 below shows the overall workflow for the FIA-MS/MS analysis.
        Prepare Extraction Solution with SIL internal standard🠦Punch 3.2 mm sample of DBS🠦Extract DBS punch with Extraction solution🠦Supernatent dried under nitrogen stream🠦Residue derivatized to butyl esters🠦Dried under stream of nitrogen🠦Residue reconstituted to solution for FIA-MS/MS
        Figure 1.Workflow for acylcarnitine analysis in dried blood spots (DBS) by FIA-MS/MS.
        Quantification of acylcarnitines using stable isotope-labeled (SIL) internal standards
        This article illustrates the utility of accurate and convenient stable isotope-labeled (SIL) internal standard solutions from Supelco for the analysis of acylcarnitine panel by FIA-MS/MS. The solution-based SIL internal standards are certified reference materials (CRMs), eliminating the need for quantitative transfer of lyophilized internal standards prior to dilution and provide accuracy and flexibility to prepare only the required amount of extraction solution with SIL internal standards.
        Acylcarnitine DBS reference samples from CDC were used as controls and two levels of control DBS samples from Recipe ClinChek with known concentrations for 13 acylcarnitines were used as samples for the analysis.
        2. Detailed Acylcarnitine FIA-MS/MS Workflow
        2.1 Acylcarnitine SIL Internal Standards
        Stable isotope-labeled (SIL) Acylcarnitine CRM solutions with 13 different heavy atom labeled acylcarnitines were used for the accurate quantification of different acylcarnitine species with the most closely related internal standard. Stable isotope-labeled CRMs in solution form offer greater flexibility, accuracy, homogeneity, and convenience in use than lyophilized materials. CRMs solution eliminates the need for quantitative transfer of lyophilized content for dilution to DBS extraction solution. Figure 2 shows the chemical structures, isotope labeling, and concentrations of the CRMs used as internal standards for the acylcarnitines analysis.
        Chemical structures of internal standards used for the acylcarnitines analysis.
        Figure 2.Chemical structures of internal standards used for the acylcarnitines analysis.
        2.2 Acylcarnitine Sample Extraction and Derivatization steps
        Table below shows the sequential steps involved in the extraction of acylcarnitines from DBS samples and derivatization into their butyl esters for enhanced sensitivity in the MS/MS.
        Extraction of DBS Punch
        Using an 1/8-inch hole puncher a DBS spot is punched out into a 1.5 mL microcentrifuge tube🠦100 µL of the extraction solution with SIL internal standards is assed to the DBG punch🠦Spin the sample tubes on a centrifuge and 95 µL of the spernatant to a new tube🠦The solution is concenrated to dryness under a stream of nitogen
        Derivatization of DBS extract
        Add 100 µL of 3 M HCI in 1-butanol and vortex🠦Incubate at 60° C for 30 minutes🠦The contents of the tube is dried under a steam of nitrogen🠦Vortex the derivatized residue methanol/water (80:20) mixture🠦Centrifuge the solution and supernatant was analyzed by FIA-MS/MS
        Figure 3. DBS sample extraction and derivatization steps
        2.3 DBS Extraction Solution Preparation
        The DBS extraction solution can be conveniently prepared by combining and diluting an appropriate amount of A-148-1ML, A-147-1ML, A-163 -1ML, A-181-1ML, and A-164-1ML in a volumetric flask with methanol (Figure 4). The concentrations of SIL acylcarnitines in the DBS extraction solution thus prepared are given in Table 1.
        StepPreparation of DBS Extraction Solution with SIL Acylcarnitine Internal Standards
        1Sonicate the SIL Acylcarnitines solution ampules at room temperature for 5 minutes and mix by vortexing
        2Using a positive displacement pipette transfer 32 µL of A-148-1ML, 30 µL of A-147-1ML, 10 µL of A-163 -1ML, 75 µL of A-181-1ML and 20 µL of A-164-1ML to a 250 mL Volumetric flask containing about 100 mL of LCMS grade methanol
        3Make up the volume of the 250 mL Volumetric flask with LCMS grade methanol and mix well. Store this extraction solution according to labeling guidance
        Schematic of CRMs solution dilution by methanol in 250 mL volumetric flask
        Figure 4.Dilution schematic to prepare DBS extraction solution
        SigmaAldrich Part. No
        SIL Carnitine
        Volume of CRM soln (µL) diluted to 250 mL
        Conc. of CRM solution
        Conc. of DBS Extraction Solution
        µg/mL
        µM
        µM
        mg/L
        A-148-1ML
        C0-Carnitine-D9
        32
        1000
        5873
        0.7518
        0.128
        A-148-1ML
        C2-Carnitine-D3
        32
        1000
        4848
        0.6206
        0.128
        A-163-1ML
        C3-Carnitine-D3
        10
        501
        2275
        0.0910
        0.020
        A-147-1ML
        C4-Carnitine-D3
        30
        100
        426.8
        0.0512
        0.012
        A-147-1ML
        iC5-Carnitine-D9
        30
        100
        393.1
        0.0472
        0.012
        A-181-1ML
        C5DC-Carnitine-D3
        75
        100
        360.6
        0.1082
        0.030
        A-147-1ML
        C6-Carnitine-D3
        30
        100
        381.2
        0.0457
        0.012
        A-147-1ML
        C8-Carnitine-D3
        30
        100
        344.3
        0.0413
        0.012
        A-147-1ML
        C10-Carnitine-D3
        30
        100
        314.0
        0.0377
        0.012
        A-147-1ML
        C12-Carnitine-D9
        30
        100
        283.6
        0.0340
        0.012
        A-147-1ML
        C14-Carnitine-D9
        30
        100
        262.7
        0.0315
        0.012
        A-164-1ML
        C16-Carnitine-D3
        20
        500
        1242
        0.0994
        0.040
        A-147-1ML
        C18-Carnitine-D3
        30
        200
        464.4
        0.0557
        0.024
        Table 1. Concentration of SIL acylcarnitines in DBS extraction solution
        2.4 Extraction and Derivatization of DBS punch
        Extraction steps of DBS punch is given below.
        StepExtraction of DBS punch
        1DBS collection card without blood was used as a blank sample and 4 levels of DBS reference standards from CDC were used as control samples. Additionally, 2 levels of DBS standards from Recipe ClinChek-Control (Part# MS10182) were also extracted as reference samples. Blank and controls were extracted as detailed in the steps below.
        2Using a 1/8 inch single hole puncher a DBS spot is punched out into a 1.5 mL microcentrifuge tube.
        3100 µL of the Extraction solution with SIL internal standards is added to the DBS punch and shaken on a rocker for 30 minutes at room temperature.
        4Spin the sample tubes on a microcentrifuge and 95 µL of the supernatant is transferred to a new microcentrifuge tube and the solution is concentrated to dryness under a stream of nitrogen.
        The table below describes the steps involved in the Derivatization of DBS extract for enhanced sensitivity in MS.
        StepDerivatization of DBS extract
        1Add 100 µL of 3M HCl in 1-butanol to the dried extract residue and vortex at 1200 rpm for 1 minute
        2Incubate at 60˚C for 30 minutes followed by spinning the sample tubes on a microcentrifuge
        3The contents of the tube are dried under a stream of nitrogen
        4Vortex the derivatized residue with 100 µL of methanol/water (80:20) mixture at 1500 rpm for 2 minutes
        5Spin the resulting solution on a microcentrifuge and 95 µL of the supernatant was transferred to an HPLC autosampler vial with vial inserts
        2.5 FIA-MS/MS conditions
        Experimental parameters for FIA-MS/MS:
        LC-MS/MS
        Column:None
        Detection:MRM Acquisition with Electrospray Ionization in Positive Mode
        Instrumentation:Agilent 1290 Infinity II coupled to Ultivo TQ
        Mobile phase A:0.1% Formic acid in LC/MS Grade Water
        Mobile phase B:0.1% Formic acid in LC/MS Grade Acetonitrile
        Injection Volume1 µL
        Analysis Time3 minutes
        HPLC Gradient conditions:
        StepTime (min)Mobile Phase A%Mobile Phase B%Flow rate
        10.0050500.05 mL/min
        22.0050500.05 mL/min
        33.0050500.15 mL/min
        MS Source Parameters:
        Source ConditionsValue (+)
        Gas Temp (°C)275
        Gas Flow (L/min)12.0
        Nebulizer (psi)45
        Sheath Gas Temperature (°C)300
        Sheath Gas Flow (L/min)10.0
        Capillary Voltage4000
        Nozzle Voltage0
        2.6 Acquisition Parameters for Derivatized Acylcarnitines
        Compound
        Analyte/ISTD
        Precursor (m/z)
        Product (m/z)
        Dwell (ms)
        Fragmentor (V)
        CE (V)
        Polarity
        BuEster C0
        Analyte
        218.2
        85
        30
        100
        27
        Positive
        BuEster C0 d9
        ISTD
        227.2
        85
        30
        100
        27
        Positive
        BuEster C2
        Analyte
        260.2
        85
        30
        100
        22
        Positive
        BuEster C2 d3
        ISTD
        263.2
        85
        30
        100
        22
        Positive
        BuEster C3:0
        Analyte
        274.2
        85
        30
        105
        22
        Positive
        BuEster C3:0 d3
        ISTD
        277.2
        85
        30
        105
        22
        Positive
        BuEster C4:0
        Analyte
        288.2
        85
        30
        115
        24
        Positive
        BuEster C4:0 d3
        ISTD
        291.2
        85
        30
        115
        24
        Positive
        BuEster C5:1
        Analyte
        300.3
        85
        30
        115
        26
        Positive
        BuEster C5:0
        Analyte
        302.2
        85
        30
        115
        26
        Positive
        BuEster C4OH
        Analyte
        304.2
        85
        30
        115
        24
        Positive
        BuEster iC5:0 d9
        ISTD
        311.3
        85
        30
        115
        26
        Positive
        BuEster C6:0
        Analyte
        316.2
        85
        30
        115
        26
        Positive
        BuEster C5OH
        Analyte
        318.3
        85
        30
        115
        26
        Positive
        BuEster C6:0 d3
        ISTD
        319.2
        85
        30
        115
        26
        Positive
        BuEster C8:0
        Analyte
        344.3
        85
        30
        125
        28
        Positive
        BuEster C8:0 d3
        ISTD
        347.3
        85
        30
        125
        28
        Positive
        BuEster C3DC
        Analyte
        360.3
        85
        30
        125
        30
        Positive
        BuEster C10:1
        Analyte
        370.3
        85
        30
        135
        28
        Positive
        BuEster C10:0
        Analyte
        372.3
        85
        30
        135
        28
        Positive
        BuEster C10:0 d3
        ISTD
        375.3
        85
        30
        135
        28
        Positive
        BuEster C5DC
        Analyte
        388.2
        85
        30
        125
        30
        Positive
        BuEster C5DC d3
        ISTD
        391.3
        85
        30
        125
        30
        Positive
        BuEster C12:1
        Analyte
        398.3
        85
        30
        135
        30
        Positive
        BuEster C12:0
        Analyte
        400.3
        85
        30
        135
        30
        Positive
        BuEster C12:0 d9
        ISTD
        409.4
        85
        30
        135
        30
        Positive
        BuEster C14:2
        Analyte
        424.3
        85
        30
        135
        32
        Positive
        BuEster C14:1
        Analyte
        426.3
        85
        30
        135
        32
        Positive
        BuEster C14:0
        Analyte
        428.4
        85
        30
        135
        32
        Positive
        BuEster C14:0 d9
        ISTD
        437.4
        85
        30
        135
        32
        Positive
        BuEster C16:1
        Analyte
        454.4
        85
        30
        135
        34
        Positive
        BuEster C16:0
        Analyte
        456.4
        85
        30
        135
        34
        Positive
        BuEster C16:0 d3
        ISTD
        459.4
        85
        30
        135
        34
        Positive
        BuEster C16OH
        Analyte
        472.4
        85
        30
        135
        34
        Positive
        BuEster C18:2
        Analyte
        480.3
        85
        30
        135
        35
        Positive
        BuEster C18:1
        Analyte
        482.4
        85
        30
        135
        35
        Positive
        BuEster C18:0
        Analyte
        484.5
        85
        30
        135
        35
        Positive
        BuEster C18:0 d3
        ISTD
        487.5
        85
        30
        135
        35
        Positive
        BuEster C18OH
        Analyte
        500.5
        85
        30
        135
        35
        Positive
        Table 2. MS/MS MRM acquisition parameters for each analyte and internal standards
        3. Acylcarnitine FIA-MS/MS Quantification and Results
        3.1 MRM Chromatogram of SIL Internal Standards
        MRM trace for butyl ester derivatized SIL acylcarnitines C0 d9, C10:0 d3, C12:0 d9, C14:0 d9, C16:0 d3, C18:0 d3, C2 d3 and C3 d3
        Figure 5.MRM trace for butyl ester derivatized SIL acylcarnitines C0 d9, C10:0 d3, C12:0 d9, C14:0 d9, C16:0 d3, C18:0 d3, C2 d3 and C3 d3
        MRM chromatograms for SIL acylcarnitines with peaks in flow injection
        Figure 6.MRM trace for butyl ester derivatized SIL acylcarnitines C4 d3, C5DC d3, C6 d3, C8:0 d3 and iC5 d9
        3.2 MRM Chromatogram of Blank and Control Injections
        Overlap chromatograms for all MRM transitions that include SIL internal standards and native acylcarnitines are shown in Figure 5. In the first overlapped MRM chromatogram, the control blank injection has a baseline signal for injection of methanol-water mixture. Whereas the second MRM overlap shows the injection of DBS extract from a CDS control sample.
        Blank (80:20 methanol/water) injections: Shows no background Interferences for all analytes and SIL Internal Standards
        Blank (80:20 methanol/water) injections: Shows no background Interferences for all analytes and SIL Internal Standards
        Chromatograms for all MRM transition with control blank injection and DBS extract from a CDS control
        Figure 7.MRM traces for all analytes and SIL internal standards with a blank injection of 80:20 methanol/water (top) and CDC Level-D DBS control (bottom)
        3.3 Quantification of Acylcarnitines and Interpretation of Results
        Acylcarnitine concentration is determined by comparing the MRM signal intensities against the know concentration of closely related SIL internal standards using the formula in the table below.
        CA = (AA x VEX x CIS) ÷ (VB x AIS)
        CA Target analyte concentration in blood (µM)
        AA Area count for analyte
        VEX Volume of Extraction solution (µL)
        CIS Concentration of SIL internal standard in Extraction solution (µM)
        VB Volume of blood in DBS punch (µL) (3.1 µL for 3.2 mm DBS punch
        AIS Area count for SIL internal standard
        Figure 8.Formula to calculate acylcarnitine concentration in DBS sample
        Accuracy was demonstrated through quantification of the four levels of control DBS samples from CDC with the known concentration of 20 acylcarnitines. The samples were analyzed (minimum of 3 replicate DBS extractions for each level) and the measured values were plotted against the actual mean concentrations of the control standards.6 As shown in Table 3, the correlation (slope) and R2 is close to 1 for all acylcarnitines, indicative of the good accuracy of the quantification. The correlations are shown graphically in Figures 9 and 10.
        In addition, two levels of control DBS samples from Recipe ClinChek with known concentration for 13 acylcarnitines were analyzed (in 6 replicate extractions for each level). As indicated in Table 4, the measured concentrations of all the acylcarnitines were within the expected control range.
        Evaluation of measured acylcarnitines concentration vs. actual concentration in CDC control DBS Samples
        Analyte
        Slope
        R2
        Intercept
        C0
        1.1
        0.9941
        -3.175
        C2
        0.8
        0.9930
        -0.873
        C3
        0.8
        0.9964
        -0.128
        C3DC
        1.0
        0.9949
        -0.062
        C4:0
        0.8
        0.9927
        0.083
        C4OH
        0.8
        0.9955
        -0.015
        C5:0
        0.9
        0.9977
        -0.035
        C5:1
        1.0
        0.9974
        -0.054
        C5DC
        0.9
        0.9968
        -0.040
        C5OH
        0.8
        0.9961
        -0.013
        C6:0
        0.8
        0.9987
        -0.009
        C8:0
        0.8
        0.9977
        -0.034
        C10:0
        0.7
        0.9982
        -0.028
        C12:0
        0.8
        0.9986
        -0.036
        C14:0
        0.8
        0.9993
        -0.025
        C14:1
        0.9
        0.9988
        -0.028
        C16:0
        0.9
        0.9982
        -0.108
        C16OH
        0.8
        0.9992
        -0.004
        C18:0
        0.9
        0.9984
        -0.056
        C18OH
        0.9
        0.9986
        -0.008
        Table 3. Correlation (slope) between the measured concentration and the actual mean concentration of acylcarnitines in DBS control standards from CDC
        Evaluation of measured acylcarnitines concentration vs. actual concentration in Recipe control DBS samples
        Acylcarnitine
        Control Level
        Average Measured Concentration (µM)
        Expected Control Range (µM)
        C0
        Level-I
        26.1
        11.7 - 40.4
        Level-II
        170
        65.9 - 198
        C2
        Level-I
        11.1
        3.37 - 15.9
        Level-II
        76.2
        29.9 - 119
        C3
        Level-I
        1.86
        1.59 - 3.30
        Level-II
        12.7
        10.2 - 19.0
        C4
        Level-I
        0.753
        0.645 - 1.34
        Level-II
        6.92
        5.90 - 11.0
        C5
        Level-I
        0.478
        0.271 - 0.713
        Level-II
        2.34
        1.31 - 3.06
        C5DC
        Level-I
        0.552
        0.462 - 1.85
        Level-II
        2.46
        1.98 - 6.82
        C6
        Level-I
        0.366
        0.260 - 0.541
        Level-II
        1.08
        0.748 - 1.39
        C8:0
        Level-I
        0.374
        0.225 - 0.675
        Level-II
        2.31
        1.39 - 3.67
        C10:0
        Level-I
        0.196
        0.112 - 0.336
        Level-II
        1.04
        0.593 - 1.56
        C12:0
        Level-I
        0.406
        0.262 - 0.544
        Level-II
        6.61
        3.89 - 7.23
        C14:0
        Level-I
        0.421
        0.270 - 0.711
        Level-II
        3.42
        2.12 - 4.94
        C16:0
        Level-I
        0.986
        0.661 - 1.98
        Level-II
        9.22
        5.82 - 15.3
        C18:0
        Level-I
        0.524
        0.327 - 0.982
        Level-II
        4.04
        2.37 - 6.26
        Table 4.Comparison of the average measured concentration of acylcarnitines against the expected concentration range in the Recipe ClinChek control DBS samples
        Correlation plots between measured and actual acylcarnitines concentration in CDC control DBS samples
        Correlation plot for the comparison of measured concentrations of acylcarnitines (C0, C2, C3, C3DC, C4, C4OH, C5:0, C5:1, C5DC) against the expected concentrations (MS/MS non-kit) in the four levels of CDC DBS controls
        Figure 9. Correlation plot for the comparison of measured concentrations of acylcarnitines (C0, C2, C3, C3DC, C4, C4OH, C5:0, C5:1, C5DC) against the expected concentrations (MS/MS non-kit) in the four levels of CDC DBS controls
        Correlation plot for the comparison of measured concentrations of acylcarnitines (C5OH, C6:0, C8:0, C10:0, C12:0, C14:0, C14:1, C16:0, C16OH, C18:0, C18OH) against the expected concentrations (MS/MS non-kit) in the four levels of CDC DBS controls
        Figure 10.Correlation plot for the comparison of measured concentrations of acylcarnitines (C5OH, C6:0, C8:0, C10:0, C12:0, C14:0, C14:1, C16:0, C16OH, C18:0, C18OH) against the expected concentrations (MS/MS non-kit) in the four levels of CDC DBS controls
        4. Conclusion: Quantitative Analysis of Acylcarnitines from Dried Blood Spots
        This study illustrates the use of stable isotope-labeled (SIL) Acylcarnitines Certified Reference Materials (CRM) for the quantitation of acylcarnitines in dried blood spots. FIA-MS/MS analysis in combination with the SIL Acylcarnitine CRM solutions from Supelco can be used effectively to accurately measure the concentration of different acylcarnitine species in dried blood spots (DBS). Due to the availability of a wide diversity of SIL acylcarnitine species from Supelco, the SIL Acylcarnitine CRM that is most chemically similar to the analyte could be chosen as the MS/MS reference for the utmost accurate quantification. The SIL CRMs in solutions offer greater flexibility, accuracy, homogeneity, and convenience than lyophilized materials. Solutions allow flexibility to prepare any volume of DBS extraction solution with the reference materials, as desired by the analyst. The CRMs in solution form eliminate the need for quantitative transfer of the lyophilized content for dilution to DBS Extraction Solution. A complete series of native Acylcarnitines Mix CRM solutions are also available from Supelco for instrument calibration, controls, and MS/MS tuning.
        Materials
        Sorry, an unexpected error has occurred
        Response not successful: Received status code 500
        References
        1.
        De Jesús VR, Chace DH, Lim TH, Mei JV, Hannon WH. 2010. Comparison of amino acids and acylcarnitines assay methods used in newborn screening assays by tandem mass spectrometry. Clinica Chimica Acta. 411(9-10):684-689. https://doi.org/10.1016/j.cca.2010.01.034
        2.
        Zytkovicz TH, Fitzgerald EF, Marsden D, Larson CA, Shih VE, Johnson DM, Strauss AW, Comeau AM, Eaton RB, Grady GF. 2001. Tandem Mass Spectrometric Analysis for Amino, Organic, and Fatty Acid Disorders in Newborn Dried Blood Spots. 47(11):1945-1955. https://doi.org/10.1093/clinchem/47.11.1945
        3.
        Chace DH, Lim T, Hansen CR, De Jesus VR, Hannon WH. 2009. Improved MS/MS analysis of succinylacetone extracted from dried blood spots when combined with amino acids and acylcarnitine butyl esters. Clinica Chimica Acta. 407(1-2):6-9. https://doi.org/10.1016/j.cca.2009.06.017
        4.
        Haynes CA, De Jesús VR. 2016. Simultaneous quantitation of hexacosanoyl lysophosphatidylcholine, amino acids, acylcarnitines, and succinylacetone during FIA?ESI?MS/MS analysis of dried blood spot extracts for newborn screening. Clinical Biochemistry. 49(1-2):161-165. https://doi.org/10.1016/j.clinbiochem.2015.09.011
        5.
        Chace DH, Kalas TA, Naylor EW. 2003. Use of Tandem Mass Spectrometry for Multianalyte Screening of Dried Blood Specimens from Newborns. 49(11):1797-1817. https://doi.org/10.1373/clinchem.2003.022178
        6.
        Newborn Screening Quality Assurance Program 2020 Quality Control Report. June 1, 2020. [Internet]. Available from: https://www.cdc.gov/labstandards/pdf/nsqap/QC_Report_S1_2020-508.pdf