MilliporeSigma
HomeWet Chemical AnalysisEMSURE® Grade Inorganic Solvents & Reagents
EMSURE® Grade Inorganic Solvents & Reagents
Introduction
EMSURE® premium grade Inorganic Reagents and Solvents are optimized for classical analytical applications. Most EMSURE® products meet and exceed ACS, Reag. Ph Eur, and ISO guidelines, ensuring compliance with international standards for regulated analyses and advanced laboratory applications. These products guarantee high quality and comprehensive specifications to minimize uncertainties and control test conditions, making them suitable for an extraordinarily wide range of applications.
A bar chart comparing the number of analytical parameters for sodium chloride for analysis provided by EMSURE® (Cat. No. 106404) versus three other brands (Brand A, Brand B, and Brand C). The leftmost bar represents the EMSURE® product, segmented into four distinct sections, each representing different levels of regulatory compliance and additional quality specifications. The bottom section, in a dotted pattern, corresponds to the parameters required by ACS (American Chemical Society). Above that, a green solid section represents additional parameters required for Reag. Ph Eur (European Pharmacopoeia Reagent). A yellow striped section signifies additional parameters meeting ISO 6353 standards. The topmost purple solid section indicates further enhanced quality parameters unique to EMSURE®. Brands A, B, and C, represented by shaded bars to the right of EMSURE®, show fewer total parameters compared to EMSURE®. Brand A has the highest number of parameters among the competing brands, followed by Brands C and B.
Key features:
  • Specified beyond Standards: Most EMSURE® products specifications meet and exceed ACS, Reag. Ph Eur, and ISO guidelines.
  • High Sensitivity: Designed for advanced analytical methods with low detection limits.
  • Rigorous Testing: Each product is tested for up to 70 parameters, ensuring exceptional purity and low limiting values.
  • Extended Impurity Profile: Detailed Certificates of Analysis accompany each product, providing extensive impurity information for analytical confidence.
Consistency and Reliability
EMSURE® products offer batch-to-batch consistency, ensuring reliable results and reducing analytical costs by minimizing repeat analyses.
A bar chart comparing the assay effective values of different batches of Acetone for analysis, EMSURE® ACS, ISO, Reag. Ph Eur. The x-axis displays batch numbers (e.g., K44105214, K44065214, K44014914) corresponding to various production lots. The y-axis represents the percentage purity, ranging from 99.0% to 100.0%.  All the purple bars indicate the effective assay values for each batch, consistently exceeding the green horizontal line representing the assay specification limit (set slightly above 99.8%). The uniform height of the bars shows minimal variation across batches, signifying high purity and consistent quality well above the required specification. This underscores the reliability and superior quality of EMSURE® acetone.
Acetone for analysis EMSURE® ACS, ISO, Reag. Ph Eur
Convenience and Safety
EMSURE® products feature innovative packaging for safe handling, storage, and transportation, protecting chemicals from contamination. EMSURE® products come in a wide selection of packaging materials and sizes. Find out more about the highlights of our packaging range:
A variety of laboratory Emsure solvent bottles, labeled with product information, placed on a white counter in a modern laboratory setting. The bottles vary in size and shape, with both amber and white bottles visible, representing different chemical solvents typically used in analytical and research labs. The background features lab equipment and shelves
Comprehensive Documentation
EMSURE® products include detailed Safety Data Sheets (SDS), product specifications, batch-specific Certificates of Analysis (CoA), and physicochemical data, facilitating regulatory compliance and reliable results. EMSURE® products are covered by our M-Clarity™ program and are allocated to a level of MQ 300 to offer even more documentation and change notification.
overlapping pages of a Certificate of Analysis (CoA) for an EMSURE® product, featuring the Merck logo prominently at the top. The document lists various analytical parameters and corresponding batch-specific values in a detailed table format. Each parameter includes measured values along with regulatory compliance limits for comparison. The impurity profile is thorough, emphasizing the high quality of EMSURE® products, which offer maximum specifications with minimal impurities.
Cost-Effective Alternatives
  • EMPARTA® Reagents: Cost-efficient inorganic reagents for routine analyses, covering essential parameters.
  • EMPLURA® Reagents: Economical choice for basic laboratory work, available in flexible packaging sizes.
Specification
Purity
Number of Specified Parameters
MQ Level
Documentation support
Regulations
EMSURE®
The most extensive specifications worldwide!
99.7–99.9%
< 70
MQ300
CoAs, SDS, BSE/TSE or AO certificates, Special documentation on request
ACS (Reag. USP)
ISO
Reag. Ph Eur
EMPARTA®
All ACS requirements.
99.0–99.5%
< 10
MQ200
CoAs, SDS, BSE/TSE or AO certificates
ACS (Reag. USP)
EMPLURA®
All basic parameters.
~ 99%
4–5
MQ200
CoAs, SDS
-
Request your Empower your Lab catalog copy to discover inorganic reagents and solvents in three quality grades for your tailored needs and explore smart solutions to enhance your lab.
Discover the complete range of EMSURE® products.
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500