LC-MS Analysis of PFAS Compounds
Petra Lewits, Product Manager Analytical Chromatography, HPLC Columns, Cory Muraco, Product Manager, Liquid Chromatography Technology
MilliporeSigma
Section Overview
Introduction
PFAS (Per- and poly-fluoroalkyl substances) are persistent, man-made organic compounds, widely found in the environment. Recent awareness has brought attention to the toxicity of these substances. The U.S. Food and Drug Administration (FDA) and the U.S. Environmental Protection Agency (EPA) have initiated actions against PFAS. For determination of PFASs, liquid chromatography–mass spectrometry (LC-MS) is a commonly used technique.
The EPA has developed, validated, and published three methods to support the analysis of 29 PFAS in drinking water, method 533, 537 and 537.1. EPA 8327 covers the analysis of selected per- and polyfluoroalkyl substances (PFAS) in prepared extracts of various matrices (e.g., waters and solids) by liquid chromatography/tandem mass spectrometry (LC-MS/MS) analysis.
The Ascentis® Express PFAS HPLC column is designed for the separation of novel and legacy short-chain and long-chain PFAS compounds containing branched and linear isomers, whilst adhering to EPA methodology requirements.
Furthermore, a specific PFAS delay column prevents background PFAS contamination from interfering with the sample results in quantitative LC-MS methods (Figure 1).
Experimental
Instrument Set-up with a Delay Column
Figure 1.LC-MS instrumental set-up for PFAS analysis & results of perfluorooctanoic acid (PFOA) (MRM 413.0>368.0) using a delay column
LC-MS Method
LC-MS/MS analysis was performed using the conditions shown in Table 1.
| LC Conditions | ||
|---|---|---|
| Analytical column: | Ascentis® Express PFAS, 100 x 2.1 mm I.D., 2.7 μm (53559-U) | |
| Delay column: | Ascentis® Express PFAS Delay, 50 x 3 mm I.D., 2.7 μm (53572-U) | |
| Mobile phase: | [A] 10 mM Ammonium acetate; [B] methanol | |
| Gradient: | Time | % B |
| 0.0 | 33.0 | |
| 18.0 | 98.0 | |
| 18.1 | 100.0 | |
| 21.0 | 100.0 | |
| 21.1 | 33.0 | |
| 26.0 | End | |
| Flow rate: | 0.4 mL/min | |
| Injection volume: | 2.0 µL | |
| Pressure: | 485 bar | |
| Temperature: | 35 °C | |
| Sample solvent: | Methanol (96%), water (4%) | |
| MS Conditions | ||
| Detection: | ESI(-) MS/MS, for transition see Table 2 | |
| LC system: | Shimadzu Nexera X2 | |
| ESI LCMS system: | Shimadzu LCMS-8040 | |
| Spray voltage: | -2.0 kV | |
| Nebulizing gas: | 2 L/min | |
| Drying gas: | 15 L/min | |
| DL temp: | 250 ˚C | |
Figure 2.PFAS analysis with Ascentis® Express PFAS column and PFAS Delay column under EPA Method 533 conditions (Peak IDs in Table 2)
Figure 3.PFAS analysis with Ascentis® Express PFAS column and PFAS Delay column under EPA Method 537.1 conditions (Peak IDs in Table 2)
Figure 4.PFAS analysis with Ascentis® Express PFAS column and PFAS Delay column under EPA Method 8327 conditions (Peak IDs in Table 2)
| EPA 533 | EPA 537.1 | EPA 8327 | |||||
|---|---|---|---|---|---|---|---|
| Compound | Transition | Peak # | tR (min) | Peak # | tR (min) | Peak # | tR (min) |
| PFBA | 213.0000>169.0000 | 1 | 1.36 | 1 | 1.36 | ||
| 4:2FTS | 229.0000>85.0000 | 2 | 1.89 | 2 | 1.89 | ||
| PFPeA | 263.0000>219.0000 | 3 | 3.22 | 3 | 3.22 | ||
| PFBS | 299.0000>80.0000 | 4 | 3.81 | 1 | 3.789 | 4 | 3.81 |
| PFHpS | 279.0000>85.0000 | 5 | 3.97 | 5 | 3.97 | ||
| PFPeS | 315.0000>135.0000 | 6 | 4.79 | 6 | 4.79 | ||
| PFMPA | 327.0000>307.0000 | 7 | 5.43 | ||||
| PFHxA | 313.0000>269.0000 | 8 | 5.68 | 2 | 5.639 | 7 | 5.68 |
| PFEESA | 349.0000>80.0000 | 9 | 6.10 | ||||
| HFPO-DA | 285.0000>169.0000 | 10 | 6.34 | 3 | 6.307 | ||
| PFHpA | 363.0000>319.0000 | 11 | 7.76 | 4 | 7.723 | 8 | 7.76 |
| PFHxS | 399.0000>80.0000 | 12 | 7.99 | 5 | 7.936 | 9 | 7.99 |
| ADONA | 377.0000>250.9000 | 13 | 8.01 | 6 | 7.978 | ||
| FOSA | 427.0000>407.0000 | 10 | 9.30 | ||||
| PFOA | 413.0000>369.0000 | 14 | 9.40 | 7 | 9.368 | 11 | 9.40 |
| PFMBA | 449.0000>80.0000 | 15 | 9.51 | ||||
| PFDS | 295.0000>201.0000 | 12 | 9.70 | ||||
| PFNA | 463.0000>419.0000 | 16 | 10.75 | 8 | 10.715 | 13 | 10.75 |
| PFOS | 499.0000>80.0000 | 17 | 10.79 | 9 | 10.762 | 14 | 10.79 |
| 9Cl-PF3ONS | 530.9000>351.0000 | 18 | 11.46 | 10 | 11.439 | ||
| PFNS | 527.0000>507.0000 | 15 | 11.84 | ||||
| PFDA | 513.0000>469.0000 | 19 | 11.89 | 11 | 11.857 | 16 | 11.89 |
| 8:2FTS | 549.0000>80.0000 | 20 | 11.90 | 17 | 11.90 | ||
| N-MeFOSAA | 570.0000>419.0000 | 12 | 12.336 | 18 | 12.37 | ||
| 6:2FTS | 498.0000>78.0000 | 21 | 12.68 | 19 | 12.68 | ||
| NFDHA | 599.0000>80.0000 | 22 | 12.85 | ||||
| PFUnA | 563.0000>519.0000 | 23 | 12.86 | 13 | 12.822 | 20 | 12.86 |
| N-EtFOSAA | 584.0000>419.0000 | 14 | 12.827 | 21 | 12.87 | ||
| 11Cl-PF3OUdS | 630.7000>451.0000 | 24 | 13.33 | 15 | 13.311 | ||
| PFDoA | 613.0000>569.0000 | 25 | 13.71 | 16 | 13.690 | 22 | 13.71 |
| PFTrDA | 663.0000>619.0000 | 17 | 14.435 | 23 | 14.45 | ||
| PFTeDA | 713.0000>669.0000 | 18 | 15.083 | 24 | 15.10 | ||
Conclusion
The application tested Ascentis® Express PFAS column has demonstrated to be a valuable tool for reliable PFAS testing. The Ascentis® Express PFAS Delay column supports this effort by efficiently separating potential PFAS background originating from the instrument from the actual sample signals.
Learn more about PFAS analysis at SigmaAldrich.com/PFAS
Discover our range of PFAS certified reference materials (CRMs) for reliable PFAS analysis and quantification. Click here to see the full list.
Certified Reference Materials (CRMs)
Response not successful: Received status code 500