MilliporeSigma
HomeWastewater & Process Water​ TestingPFAS Testing Methods and Guidance for Sample Filtration
PFAS Testing Methods and Guidance for Sample Filtration
Filtration Product Selection Table for PFAS Testing
Filter Format
Membrane
Pore Size (µm)
Diameter (mm)
Product
PFAS Validated Syringe Filter*
  • Validated for 42 PFAS compounds
  • For sample filtration
Millipore PES Express®
0.22
33
SLGP033N
Syringe Filter
  • For sample filtration
Millipore PES Express®
0.45
33
SLHP033N
Nylon
0.20
33 
SLGN033
Nylon with glass fiber prefilter 
0.20
25
SLGNM25
Cut Disc Membrane Filter
  • For sample, mobile phase, and buffer filtration
Hydrophilic and hydrophobic polypropylene**
0.20, 0.45
13, 25, 47, 90
Polypropylene membrane filters
Mixed cellulose esters (MCE)
0.22, 0.45, 0.8, 5.0
13, 25, 47, 90
MCE membrane filters
Cut Disc Membrane Filter
  • For particulate fraction from stationary sources or stack testing for air emissions
Glass fiber filter without binder
0.7
90
AP4009000
Table 1.Millex® syringe filter devices and cut disc membrane filters appropriate for PFAS testing-related sample preparation and mobile phase filtration. Recommendations are based on testing water samples using modified EPA 537.1 and methanol samples with modified EPA 1633.
*PFAS validated Millex® syringe filters have been tested for 42 PFAS compounds to ensure levels below established reporting limits. For more information, refer to the User Guide.
**Hydrophilic polypropylene can be used for sample/mobile phase filtration of aqueous solvents. Hydrophobic polypropylene is recommended for sample/mobile phase filtration of organic solvents.
What are Poly- and Perfluoroalkyl Substances (PFAS)?
PFAS are poly- and perfluoroalkyl substances known as “forever chemicals” and comprise a group of over 4,000 varieties of long- and short-chain perfluorinated compounds.1 PFAS are used in a variety of industries for their excellent oil-, water-, temperature-, chemical-, and fire-resistant properties and noted for their use in polymerization reactions of fluoropolymers such as Teflon® by companies such as 3M and Dupont. Products that contain PFAS and related compounds are ubiquitous in industrial and consumer products, including product packaging, cosmetics, non-stick cookware, stain repellents, polishes, paints, coatings, and firefighting foams.
The excellent properties and broad use of PFAS have led to the persistent accumulation of these man-made chemicals in environmental and biological matrices, recently linked to liver damage, cancer, weakened immune system, and high cholesterol in humans.1-3
Requirement for Filtration in PFAS Methods
In response, agencies in the US and Europe have taken regulatory action. The Stockholm Convention proposed regulations for two of the most common PFAS compounds — perfluorooctanoic acid (PFOA) and perfluorooctanesulfonic acid (PFOS) — with certain exemptions, effective in 2020. The US Environmental Protection Agency (EPA) published an Action Plan in 2019 followed by recommendations for testing water matrices for PFAS compounds under the Safe Drinking Water Act in early 2020, with a drinking water advisory concentration of 70 parts per trillion (ppt). In October 2021, the EPA published its PFAS Strategic Roadmap, which outlines their extensive approach to addressing PFAS from 2021 through 2024. Most recently, the EPA released drinking water advisories for four PFAS compounds (PFOA, PFOS, Hexafluoropropylene Oxide (HFPO) dimer acid and its ammonium salt, and PFBS and its potassium salt). The European Union (EU) drinking water directive, which includes a limit of 0.5 µg/L for all PFAS, took effect in January 2021. Additionally, the European Chemicals Agency (ECHA) submitted a restriction proposal in January 2022 for PFAS in firefighting foams, with several other proposals expected through 2023. Additional PFAS substances are on the list for evaluation under REACH (Registration, Evaluation, Authorisation and Restriction of Chemicals). In response to rapidly evolving regulatory proposals and actions, academic and industrial testing labs have developed analytical methods for testing and monitoring PFAS in a variety of matrices, such as those listed in Table 2. These regulations are important to understanding the extent of human exposure and environmental contamination to inform future remediation efforts.
MethodUpdate/RevisionMatrix/MatricesSample preparationAnalytical method
EPA 537.1
Jun. 2020
Drinking water
SPE
LC-MS/MS
EPA 533
Dec. 2019
Drinking water
SPE
LC-MS/MS
SW-846 Method 8327*
Jul. 2021
Non-potable groundwater, surface water, wastewater
SPE, filtration
LC-MS/MS
ASTM D7968-17a
Sep. 2017
Environmental solids
Solvent extraction, filtration
LC-MS/MS
ASTM D7979-19
Sep. 2021
Water matrix (no drinking water)
Solvent extraction, filtration
LC-MS/MS
ISO 21675
Oct. 2019
Drinking, natural and wastewater
SPE, filtration as needed
LC-MS/MS
FDA C-010.02
Dec. 2021
Foods
QuEChERS, SPE, filtration
LC-MS/MS
OTM-45
Jan. 2021
Air emissions (stationary sources)
Sampling train: filtration, impingers
LC-MS/MS
EPA 1633
Feb. 2022
Aqueous, soil, biosolids, sediment, tissue
SPE, filtration
LC-MS/MS
EPA Draft 1621*
Apr. 2022
Aqueous matrices
TSS, GAC column cleanup
CIC
*Screening method
Abbreviations: SPE = solid phase extraction; TSS = total suspended solids determination; GAC = granular activated carbon; CIC = combustion ion chromatography
Table 2.Selected analytical methods for the detection of PFAS compounds in various matrices.
Concerns about PFAS Contamination from Sample Collection and Preparation Materials
In recent studies, there has been concern about adding PFAS contamination to samples from a variety of sources including collection bottles, solvents, storage vials, tubing components, and any other plastic that contacts the sample. This includes membrane filters, filter holders, and syringe filter housing used for clearing particulates from sample matrices. Some filters may show trace amounts of contamination that may interfere with LC-MS/MS detection of PFAS and resulting data, especially among increasing sensitivity requirements.6 Another concern specifically for consumables is the adsorption of PFAS compounds, such as onto filtration media or SPE sorbent. For filter devices, this is dependent on many attributes - most importantly the filter type, the solvent being filtered, and the type of PFAS molecule.4-5 For example, in some cases, both contamination and sorption onto filtration media can be reduced by washing with methanol.6-7
Sample Filtration in PFAS Testing
In all analytical methods, sample preparation should be carefully considered. However, in PFAS workflows, additional factors could complicate PFAS analyses downstream. These include potential PFAS contamination from filters or other consumables that contact the samples and adsorption of PFAS compounds onto consumables, leading to loss of recovery. Thus, we tested PES, nylon, and nylon-HPF syringe filter devices within PFAS detection workflows EPA 537.1 and EPA 1633 to determine levels of contamination resulting from membrane extractables. We also tested mixed cellulose ester (MCE; 0.22, 0.45, 0.8, 5.0 µm) and polypropylene cut disc membrane filters (0.2 µm and 0.45 µm hydrophilic and hydrophobic polypropylene for EPA 537.1 and 0.2 µm hydrophilic polypropylene for EPA 1633). We found that none of the filters had any detectable levels of PFAS contamination above the reporting limits (RL). Adsorption of internal standards leading to some loss of recovery occurred primarily for nylon and hydrophobic polypropylene membrane materials, which varied according to PFAS type, chain length, and filtrate material (methanol vs. water). Filtration in methanol demonstrated better recoveries of the same standards for nylon. This supports the suggestion that rinsing with methanol can reduce binding of PFAS compounds to filter materials. Hydrophilic polypropylene performed similar in both methanol and water.
Therefore, when filtration of higher particulate samples is needed in a PFAS workflow, PES, nylon, and nylon-HPF Millex® syringe filters, as well as MCE and polypropylene cut disc membrane filters, provide a suitable option.
Recommended Syringe Filters
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Recommended Membrane Filters
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Recommended Filter Holders for Membrane Filters
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Alternative diameter PES Millex® syringe filters and Millipore® cut disc membranes are also available.
References
1.
Kwiatkowski CF, Andrews DQ, Birnbaum LS, Bruton TA, DeWitt JC, Knappe DRU, Maffini MV, Miller MF, Pelch KE, Reade A, et al. 2020. Scientific Basis for Managing PFAS as a Chemical Class. Environ. Sci. Technol. Lett.. 7(8):532-543. https://doi.org/10.1021/acs.estlett.0c00255
2.
Winchell LJ, Wells MJ, Ross JJ, Fonoll X, Norton JW, Kuplicki S, Khan M, Bell KY. 2021. Analyses of per- and polyfluoroalkyl substances (PFAS) through the urban water cycle: Toward achieving an integrated analytical workflow across aqueous, solid, and gaseous matrices in water and wastewater treatment. Science of The Total Environment. 774145257. https://doi.org/10.1016/j.scitotenv.2021.145257
3.
Pérez F, Nadal M, Navarro-Ortega A, Fàbrega F, Domingo JL, Barceló D, Farré M. 2013. Accumulation of perfluoroalkyl substances in human tissues. Environment International. 59354-362. https://doi.org/10.1016/j.envint.2013.06.004
4.
Lath S, Knight ER, Navarro DA, Kookana RS, McLaughlin MJ. 2019. Sorption of PFOA onto different laboratory materials: Filter membranes and centrifuge tubes. Chemosphere. 222671-678. https://doi.org/10.1016/j.chemosphere.2019.01.096
5.
Labadie P, Chevreuil M. 2011. Biogeochemical dynamics of perfluorinated alkyl acids and sulfonates in the River Seine (Paris, France) under contrasting hydrological conditions. Environmental Pollution. 159(12):3634-3639. https://doi.org/10.1016/j.envpol.2011.07.028
6.
So MK, Taniyasu S, Lam PKS, Zheng GJ, Giesy JP, Yamashita N. 2006. Alkaline Digestion and Solid Phase Extraction Method for Perfluorinated Compounds in Mussels and Oysters from South China and Japan. Arch Environ Contam Toxicol. 50(2):240-248. https://doi.org/10.1007/s00244-005-7058-x
7.
Yamashita N, Kannan K, Taniyasu S, Horii Y, Petrick G, Gamo T. 2005. A global survey of perfluorinated acids in oceans. Marine Pollution Bulletin. 51(8-12):658-668. https://doi.org/10.1016/j.marpolbul.2005.04.026