MilliporeSigma
HomeCustom RNA Oligos
Custom RNA Oligos
Standard quantities of single-stranded RNA Oligos for various applications.
Custom RNA Oligos Specifications
Scales
  • 0.05, 0.2, 1, 10 & 15 µmol
  • For larger quantities & in-vivo-grade material, see our Custom iScale Oligos product
Purification
  • Desalt, Cartridge, HPLC or PAGE
  • Recommend purification for sequences >30 bases
Sequence Length
  • 7 to 45 bases (simplexes)
  • Inquire for feasibility over 45 bases
Modifications
  • Amino-Modifier C6, Amino-Modifier C7, Biotin, Cyanine 3, Cyanine 5, Cyanine 5.5, 6-FAM™, HEX™, TET™, Flipped Base, 2’-Fluoro, Inverted Abasic, 2’-OMe RNA, Phosphate, Thiol-Modifier-C6 S-S & Thiol-Modifier-C3 S-S
  • Locked Nucleic Acid available
  • Inquire for feasibility of Locked Nucleic Acid & other modifications
Quality Control
  • 100% mass spectrometry
Format
  • Supplied dry in tubes
  • Pooling & aliquoting available (inquire for feasibility)
Minimum Yield Guarantees
Non-Modified RNA Oligos (O.D./µg)*
Scale (µmole)DesaltCartridgeHPLCPAGE
0.055/1501/301/300.5/15
0.212/3603/902.5/751/30
1.040/1,20012/36013/3905/150
*Guarantees are for 20mers or longer. Oligos shorter than 20 bases will have fewer ODs. Microgram amounts are calculated based on a 30 μg/OD value. If the μg/OD value is less than 30, the reported μg value will be proportionally less. Phosphorothioated yields are 70% of the above stated values.
Modified RNA Oligos (O.D./µg)*
Scale (µmole)DesaltCartridgeHPLCPAGE
0.052/600.4/120.4/120.2/6
0.25/1501/301/300.4/12
1.016/4805/1505/1502/60
*Guarantees are for 20mers or longer. Oligos shorter than 20 bases will have fewer ODs. Microgram amounts are calculated based on a 30 μg/OD value. If the μg/OD value is less than 30, the reported μg value will be proportionally less. Phosphorothioated yields are 70% of the above stated values. Post-synthesis modifications may yield 50% less than the above stated values.
Shipping Schedule
Scale (µmole)DesaltCartridgeHPLCPAGE
0.05, 0.2, and 1.05 days5 days6 days8 days
For anything marked as ‘Inquire’ or if you have needs that are different from the other general specifications presented, please send a request to [email protected].
Sorry, an unexpected error has occurred
Response not successful: Received status code 500