MilliporeSigma
HomePathogen & Spoilage Testing1-2 Test® for Salmonella
1-2 Test® for Salmonella
The 1-2 Test® for Salmonella is an antibody based assay for the detection of Salmonella in food and environmental samples. The assay incorporates immunodiffusion technology to provide accurate and rapid results following a simple 1 or 2 step enrichment protocol. If present, Salmonella move through the motility media as Salmonella-specific antibodies diffuse into the media from the opposite direction. The antibodies bind to the target Salmonella antigen to form a visually apparent immunoband indicating a positive test result. As an AOAC Official Method of Analysis, the 1-2 Test is one of the simplest rapid methods available for the detection of Salmonella.
1-2 Test® for Salmonella
The First Widely Accepted Single-Use Salmonella Test
The 1-2 Test is a patented, single-use test for Salmonella used by analysts worldwide. It combines proprietary science with practical application, resulting in proven performance and ease of use.
Accurate, Reliable and Validated Results
Extensive multi-laboratory and field validation studies have shown that the 1-2 Test is highly accurate and specific. The proprietary 1-2 Test antibody captures a wide range of Salmonella, without reacting with most common cross-reactors.
One Test for all of Your Samples
The single-use format of the 1-2 Test provides the flexibility to run any number of tests depending on your testing needs. The 1-2 Test is suitable for testing all food products, food ingredients and environmental samples.
Key Tests/Pathogens: Salmonella
Validations: AOAC OMA 989.13
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500