Fortfahren mit
BR782801
BRAND® insert strips for 24 x 6-well BRANDplates®
Inlet Opening System, PC membrane, size 0.4 μm
Synonym(e):
BRANDplates®, Cell culture, Insert Strips, Insert System, Inserts, Tissue culture
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
12 ea
Warenkorb auf Verfügbarkeit prüfen
€ 346,00
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41122107
Material:
PC membrane
Pore size:
0.4 μm
Sterility:
sterile
Binding type:
non-treated surface
€ 346,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
PC membrane
description
cellGrade™ plus
sterility
sterile
manufacturer/tradename
BRAND 782801
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped), pkg of (individually wrapped)
size
0.4 μm
pore size
0.4 μm
binding type
non-treated surface
General description
Innovative cell culture plates and inserts for manual and automatic cell and tissue culture. The BRANDplates® Insert System, developed in conjunction with the Fraunhofer company, offers an innovative expansion to BRAND′s existing cell culture product range. It consists of two different cell culture plates and two designs of cell culture inserts that can be combined in different configurations.Insert specification:4x insert strips, can be divided into up to 4 individual inserts - hanging position in wellHanging inserts – distance between the well base and insert membrane is 4 mmIndividually packaged strips inserted in thermoformed beakers with 6 strips each in the 6-well cell culture platecellGrade™ plus (hydrophilic cell culture-) surface.Smooth wall inserts: For standard applications like secretion studies, co-cultivations, chemotaxis tests, etc.PS cellGrade™ plus surface, sterile, strip of 4 inserts (divisible).BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
BR782821BR782870BR782871
description
Inlet Opening System, PC membrane, size 0.4 μm
description
Inlet Opening System, PC membrane, pore size 1 μm
description
PET membrane, pore size 8 μm
description
Inlet Opening System, PET membrane, pore size 8 μm
pore size
0.4 μm
pore size
1 μm
pore size
8 μm
pore size
8 μm
manufacturer/tradename
BRAND 782801
manufacturer/tradename
BRAND 782821
manufacturer/tradename
BRAND 782870
manufacturer/tradename
BRAND 782871
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
binding type
non-treated surface
binding type
-
binding type
non-treated surface
binding type
non-treated surface
material
PC membrane
material
PC membrane
material
PET membrane
material
PET membrane
size
0.4 μm
size
6 well
size
6 well
size
6 well
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)