Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
12 ea | Warenkorb auf Verfügbarkeit prüfen | € 346,00 |
Über diesen Artikel
€ 346,00
material
PC membrane
description
cellGrade™ plus
sterility
sterile
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
manufacturer/tradename
BRAND 782821
size
6 well
pore size
1 μm
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Inlet Opening System, PC membrane, pore size 1 μm | description PET membrane, pore size 8 μm | description Inlet Opening System, PC membrane, size 0.4 μm | description Inlet Opening System, PET membrane, pore size 0.4 μm |
| pore size 1 μm | pore size 8 μm | pore size 0.4 μm | pore size 0.4 μm |
| manufacturer/tradename BRAND 782821 | manufacturer/tradename BRAND 782870 | manufacturer/tradename BRAND 782801 | manufacturer/tradename BRAND 782811 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| material PC membrane | material PET membrane | material PC membrane | material PET membrane |
| size 6 well | size 6 well | size 0.4 μm | size 6 well |
| packaging pack of 12 ea (strips of 4 inserts, individually wrapped) | packaging pack of 12 ea (strips of 4 inserts, individually wrapped) | packaging pack of 12 ea (strips of 4 inserts, individually wrapped), pkg of (individually wrapped) | packaging pack of 12 ea (strips of 4 inserts, individually wrapped) |
Still not finding the right product?
Explore all of our products under BRAND® insert strips for 24 x 6-well BRANDplates®
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?